Merck
  • Mitophagy antagonism by ZIKV reveals Ajuba as a regulator of PINK1 signaling, PKR-dependent inflammation, and viral invasion of tissues.
Mitophagy antagonism by ZIKV reveals Ajuba as a regulator of PINK1 signaling, PKR-dependent inflammation, and viral invasion of tissues.
Cell reports (2021-10-28)
Sanket S Ponia, Shelly J Robertson, Kristin L McNally, Gayatri Subramanian, Gail L Sturdevant, Matthew Lewis, Forrest Jessop, Catherine Kendall, Dylan Gallegos, Arielle Hay, Cindi Schwartz, Rebecca Rosenke, Greg Saturday, Catherine M Bosio, Craig Martens, Sonja M Best
ABSTRACT
Dysregulated inflammation dominated by chemokine expression is a key feature of disease following infection with the globally important human pathogens Zika virus (ZIKV) and dengue virus, but a mechanistic understanding of how pro-inflammatory responses are initiated is lacking. Mitophagy is a quality-control mechanism that regulates innate immune signaling and cytokine production through selective degradation of damaged mitochondria. Here, we demonstrate that ZIKV nonstructural protein 5 (NS5) antagonizes mitophagy by binding to the host protein Ajuba and preventing its translocation to depolarized mitochondria where it is required for PINK1 activation and downstream signaling. Consequent mitophagy suppression amplifies the production of pro-inflammatory chemokines through protein kinase R (PKR) sensing of mitochondrial RNA. In Ajuba-/- mice, ZIKV induces early expression of pro-inflammatory chemokines associated with significantly enhanced dissemination to tissues. This work identifies Ajuba as a critical regulator of mitophagy and demonstrates a role for mitophagy in limiting systemic inflammation following infection by globally important human viruses.
Sigma-Aldrich
Anti-phospho-PKR (Thr451) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
Epoxomicin, ≥95% (HPLC), solid
Sigma-Aldrich
Oligomycin A, ≥99% (HPLC)
Sigma-Aldrich
ISRIB, ≥98% (HPLC)
Sigma-Aldrich
Anti-phospho-Ubiquitin (Ser65), from rabbit, purified by affinity chromatography
Sigma-Aldrich
Rabbit Anti-Chicken IgY Antibody, HRP conjugate, Upstate®, from rabbit
Sigma-Aldrich
Tunicamycin Ready Made Solution, 5 mg/mL in DMSO, from Streptomyces sp.
Sigma-Aldrich
Anti-TOMM20 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
PKR Inhibitor, The PKR Inhibitor, also referenced under CAS 608512-97-6, controls the biological activity of PKR. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
PKR Inhibitor, Negative Control, The PKR Inhibitor, Negative Control, also referenced under CAS 852547-30-9, controls the biological activity of PKR. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.