Skip to Content
Merck
All Photos(4)

Key Documents

ABS1513-I

Sigma-Aldrich

Anti-phospho-Ubiquitin (Ser65)

from rabbit, purified by affinity chromatography

Synonym(s):

Ubiqutin, Ser65 phosphorylated, Ub, Ser65 phosphorylated

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), guinea pig (based on 100% sequence homology), rat (based on 100% sequence homology), horse (based on 100% sequence homology), sheep (based on 100% sequence homology), bovine (based on 100% sequence homology), hamster (based on 100% sequence homology), nonhuman primates (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

phosphorylation (pSer65)

Gene Information

human ... UBB(7314)

General description

Ubiqutin (Ub) is initially produced as a 229-a.a. Polyubiquitin-B (UniProt P0CG47) precursor protein encoded by the UBB gene (Gene ID 7314) or a 685-a.a. Polyubiquitin-C precursor protein (UniProt P0CG48) encoded by the UBC gene (Gene ID 7316) in human. The C-termini of Polyubiquitin-B (Cys229) and Polyubiquitin-C (Val685) are post-translationally removed, followed by additional posttranslational cleavages to yield multiple copies of idential 76-amino acid ubiquitin (also known as high mobility group nonhistone protein HMG-20) molecules (three copies per Polyubiquitin-B and nine copies per Polyubiquitin-C). Covalent modifications of target proteins via different Ub Lys residues are well documented events during cellular DNA repair, ER-associated degradation (ERAD), transcription activation, lysosomal and proteasomal degradation. PINK1 (PTEN induced putative kinase 1) is a Ser/Thr kinase that specifically accumulates in depolarized mitochondria, where it acts as a positive regulator of parkin (Park2) E3 ubiquitin (Ub) ligase activity by phosphorylating Ub at Ser65 as well as the corresponding Ser residue in parkin N-terminal Ub-like (UBL) domain. Phosphorylated Ub interacts with phosphorylated parkin at its GINGO0 domain in an allosteric manner, inducing a conformation change and exposing parkin RING2 domain catalytic cysteine for full ligase activity. NMR based conformation study reveals that Ser65-phosphorylated Ub exists in two conformations in solution. The major conformation displays altered surface properties, while the second phosphoUb conformation exhibits a retracted C-terminal tail by two residues into the Ub core. Ub Ser65 phosphorylation has little effect toward E1-mediated E2 charging and mainly affects the discharging of E2 enzymes to form polyUb chains. In addition, the majority of deubiquitinases (DUBs), including USP8, USP15 and USP30, are shown to exhibit impaired activity against Lys63-linked poly-phosphoUb.

Specificity

Expected to react with a broad-range of species based on 100% sequence homology.
This polyclonal antibody detects only Ser65-phosphorylated ubiquitin (Ub) and ubiquitinated proteins (including oligoUb and polyUb) containing Ser65-phosphorylated ubiquitin. No reactivity toward Ub or ubiquitinated proteins in the absence of Ub Ser65 phosphorylation.

Immunogen

KLH-conjugated linear peptide corresponding to the ubiquitin target region sequence containing phosphorylated Ser65.

Application

Detect ubiquitin Ser65 phosphorylation using this rabbit polyclonal Anti-phospho-Ubiquitin (Ser65), Cat. No. ABS1513-I, validated for use in Immunocytochemistry and Western Blotting.
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected CCCP (Cat. No. 215911) treatment-induced ubiquitin Ser65 phosphorylation in PINK1-expressing HeLa cells.

Western Blotting Analysis: A 1:5,000 dilution from a representative lot detected GST-PINK1-catalyzed Ser65 phosphorylation of recombinant Ubiquitin.

Immunocytochemistry Analysis: A 1:200 dilution from a representative lot detected phospho-Ubiquitin (Ser65) in PINK1 expressing HeLa cells without and with CCCP treatment. (Courtesy of Dr. Noriyuki Matsuda).

Quality

Evaluated by Western Blotting in PINK1-expressing HeLa cells.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected cellular ubiquitin Ser65 phosphorylation induction in 10 µg of lysate from PINK1-expressing HeLa cells upon CCCP (Cat. No. 215911) treatment.

Target description

~17/8 and ~45-250 kDa observed. 8.565/17.112/25.66/34.21 kDa (mono-/di/tri-/tetra-ubiquitin) calculated. Bands ~45-250 kDa represent ubiquitinated proteins containing ser65-phosphorylated Ub. Uncharacterized bands may be observed in some lysate(s).

Linkage

Replaces: ABS1513

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nan Wang et al.
Experimental and therapeutic medicine, 22(5), 1246-1246 (2021-09-21)
Glioma is a common malignant tumor of the central nervous system, accounting for ~50% of intracranial tumors. The current standard therapy for glioma is surgical resection followed by postoperative adjuvant radiotherapy and temozolomide (TMZ) chemotherapy. However, resistance to TMZ is
Sihang Yu et al.
International journal of molecular sciences, 22(8) (2021-05-01)
The study of cisplatin sensitivity is the key to the development of ovarian cancer treatment strategies. Mitochondria are one of the main targets of cisplatin, its self-clearing ability plays an important role in determining the fate of ovarian cancer cells.
Nan Wang et al.
Journal of cellular and molecular medicine, 26(3), 893-912 (2021-12-30)
Glioblastoma multiforme (GBM) is a primary tumour of the central nervous system (CNS) that exhibits the highest degree of malignancy. Radiotherapy and chemotherapy are essential to prolong the survival time of patients. However, clinical work has demonstrated that sensitivity of
Guillermo López-Doménech et al.
The EMBO journal, 40(14), e100715-e100715 (2021-06-22)
Clearance of mitochondria following damage is critical for neuronal homeostasis. Here, we investigate the role of Miro proteins in mitochondrial turnover by the PINK1/Parkin mitochondrial quality control system in vitro and in vivo. We find that upon mitochondrial damage, Miro
Stefanie Marek-Iannucci et al.
JCI insight, 6(18) (2021-08-18)
Kawasaki disease (KD) is the leading cause of acquired heart disease among children. Murine and human data suggest that the NLRP3-IL-1β pathway is the main driver of KD pathophysiology. NLRP3 can be activated during defective autophagy/mitophagy. We used the Lactobacillus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service