Skip to Content
Merck
All Photos(1)

Key Documents

SML1287

Sigma-Aldrich

Tunicamycin

from Streptomyces sp., ≥98% (HPLC), DMSO solution, N-acetylglucosamine transferase inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H60N4O16
CAS Number:
Molecular Weight:
816.89
EC Number:
UNSPSC Code:
51111800
NACRES:
NA.77

Product Name

Tunicamycin Ready Made Solution, 5 mg/mL in DMSO, from Streptomyces sp.

biological source

Streptomyces sp.

Quality Level

form

DMSO solution

concentration

5 mg/mL in DMSO

storage temp.

2-8°C

SMILES string

N4(C=CC(=O)NC4=O)[C@@H]1O[C@@H]([C@H]([C@H]1O)O)C(O)C[C@H]2O[C@@H]([C@@H]([C@H]([C@H]2O)O)NC(=O)\C=C\CC(C)C)O[C@H]3O[C@@H]([C@H]([C@@H]([C@H]3NC(=O)C)O)O)CO

InChI

1S/C30H46N4O16/c1-11(2)5-4-6-16(38)32-19-23(43)20(40)14(47-29(19)50-28-18(31-12(3)36)22(42)21(41)15(10-35)48-28)9-13(37)26-24(44)25(45)27(49-26)34-8-7-17(39)33-30(34)46/h4,6-8,11,13-15,18-29,35,37,40-45H,5,9-10H2,1-3H3,(H,31,36)(H,32,38)(H,33,39,46)/b6-4+/t13?,14-,15-,18-,19-,20+,21-,22-,23-,24+,25-,26-,27-,28-,29-/m1/s1

InChI key

ZHSGGJXRNHWHRS-PEALBESXSA-N

General description

Tunicamycin Ready Made Solution contains homologues A,B,C, and D. The composition may vary from lot to lot. Actual content given on label.
Tunicamycin was first isolated from Streptomyces lysosuperificus based on its antiviral activity. It contains N-acetylglucosamine and inhibits glycoprotein biosynthesis, including virus membrane glycoproteins.

Application

Tunicamycin has been used to study the effect of N-linked glycosylation of human proton-coupled folate transporter (HsPCFT) in HeLa cells. Tunicamycin has also been used to study the functional effects of coxsackievirus and adenovirus receptor (CAR) glycosylation in COS-7 cells.

Biochem/physiol Actions

Tunicamycin is an antibacterial and antifungal.
Tunicamycin is an antibacterial and antifungal. Tunicamycin blocks the formation of protein N-glycosidic linkages by inhibiting the transfer of N-acetylglucosamine 1-phosphate to dolichol monophosphate. Tunicamycin inhibits bacterial and eukaryote N-acetylglucosamine transferases and prevents formation of N-acetylglucosamine lipid intermediates.
Tunicamycin plays an important role in the antitumor activity of trastuzumab on breast cancer in vitro and in vivo. In human cell lines, tunicamycin enhances cytotoxicity stimulated by cisplatin. It prevents angiogenesis in nude mice by reducing the expression of vascular endothelial growth factor (VEGF).

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Laïla Giordano et al.
Molecular plant pathology, 23(12), 1783-1791 (2022-09-15)
The phytohormone abscisic acid (ABA) regulates cell growth and plant development, and contributes to defence responses to pathogens. We previously showed that the Arabidopsis malectin-like domain leucine-rich repeat receptor-like kinase (MLD-LRR-RLK) impaired oomycete susceptibility 1 (IOS1) attenuates ABA signalling during
The specific site of tunicamycin inhibition in the formation of dolichol-bound N-acetylglucosamine derivatives.
L Lehle et al.
FEBS letters, 72(1), 167-170 (1976-11-15)
Tunicamycin enhances the antitumor activity of trastuzumab on breast cancer in vitro and in vivo.
Han X, et al.
Oncotarget, 6(36), 38912?38925-38912?38925 (2015)
Sjors M Kas et al.
Scientific reports, 13(1), 17648-17648 (2023-10-18)
Congenital disorders of glycosylation (CDG) are rare genetic disorders with a spectrum of clinical manifestations caused by abnormal N-glycosylation of secreted and cell surface proteins. Over 130 genes are implicated and next generation sequencing further identifies potential disease drivers in
Wen-An Wang et al.
Journal of cell science, 136(6) (2023-02-23)
The coronavirus SARS-CoV-2, the agent of the deadly COVID-19 pandemic, is an enveloped virus propagating within the endocytic and secretory organelles of host mammalian cells. Enveloped viruses modify the ionic homeostasis of organelles to render their intra-luminal milieu permissive for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service