Skip to Content
Merck
All Photos(1)

Key Documents

47135

Supelco

Aspartame

analytical standard

Synonym(s):

Asp-Phe methyl ester, N-(L-α-Aspartyl)-L-phenylalanine methyl ester, Asp-Phe-OMe, Aspartame

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH2CH(NH2)CONHCH(CH2C6H5)COOCH3
CAS Number:
Molecular Weight:
294.30
Beilstein:
2223850
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:
E Number:
E951

grade

analytical standard

Quality Level

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

COC(=O)[C@H](Cc1ccccc1)NC(=O)[C@@H](N)CC(O)=O

InChI

1S/C14H18N2O5/c1-21-14(20)11(7-9-5-3-2-4-6-9)16-13(19)10(15)8-12(17)18/h2-6,10-11H,7-8,15H2,1H3,(H,16,19)(H,17,18)/t10-,11-/m0/s1

InChI key

IAOZJIPTCAWIRG-QWRGUYRKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Maersk et al.
The American journal of clinical nutrition, 95(2), 283-289 (2011-12-30)
The consumption of sucrose-sweetened soft drinks (SSSDs) has been associated with obesity, the metabolic syndrome, and cardiovascular disorders in observational and short-term intervention studies. Too few long-term intervention studies in humans have examined the effects of soft drinks. We compared
Jean-Daniel Berset et al.
Chemosphere, 88(5), 563-569 (2012-04-17)
A HPLC-MS/MS method is presented for the simultaneous determination of frequently used artificial sweeteners (ASs) and the main metabolite of aspartame (ASP), diketopiperazine (DKP), in environmental water samples using the direct-injection (DI) technique, thereby achieving limits of quantification (LOQ) of
Eva S Schernhammer et al.
The American journal of clinical nutrition, 96(6), 1419-1428 (2012-10-26)
Despite safety reports of the artificial sweetener aspartame, health-related concerns remain. We prospectively evaluated whether the consumption of aspartame- and sugar-containing soda is associated with risk of hematopoetic cancers. We repeatedly assessed diet in the Nurses' Health Study (NHS) and
B A Magnuson et al.
Critical reviews in toxicology, 37(8), 629-727 (2007-09-11)
Aspartame is a methyl ester of a dipeptide used as a synthetic nonnutritive sweetener in over 90 countries worldwide in over 6000 products. The purpose of this investigation was to review the scientific literature on the absorption and metabolism, the
Katsuyoshi Masuda et al.
PloS one, 7(4), e35380-e35380 (2012-04-27)
One of the most distinctive features of human sweet taste perception is its broad tuning to chemically diverse compounds ranging from low-molecular-weight sweeteners to sweet-tasting proteins. Many reports suggest that the human sweet taste receptor (hT1R2-hT1R3), a heteromeric complex composed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service