Skip to Content
Merck
All Photos(1)

Documents

A1320000

Aspartame

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Asp-Phe methyl ester, N-(L-α-Aspartyl)-L-phenylalanine methyl ester, Asp-Phe-OMe, Aspartame

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH2CH(NH2)CONHCH(CH2C6H5)COOCH3
CAS Number:
Molecular Weight:
294.30
Beilstein:
2223850
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

Agency

EP Reference Standard

API family

aspartame

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COC(=O)[C@H](Cc1ccccc1)NC(=O)[C@@H](N)CC(O)=O

InChI

1S/C14H18N2O5/c1-21-14(20)11(7-9-5-3-2-4-6-9)16-13(19)10(15)8-12(17)18/h2-6,10-11H,7-8,15H2,1H3,(H,16,19)(H,17,18)/t10-,11-/m0/s1

InChI key

IAOZJIPTCAWIRG-QWRGUYRKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Aspartame EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jean-Daniel Berset et al.
Chemosphere, 88(5), 563-569 (2012-04-17)
A HPLC-MS/MS method is presented for the simultaneous determination of frequently used artificial sweeteners (ASs) and the main metabolite of aspartame (ASP), diketopiperazine (DKP), in environmental water samples using the direct-injection (DI) technique, thereby achieving limits of quantification (LOQ) of
Eva S Schernhammer et al.
The American journal of clinical nutrition, 96(6), 1419-1428 (2012-10-26)
Despite safety reports of the artificial sweetener aspartame, health-related concerns remain. We prospectively evaluated whether the consumption of aspartame- and sugar-containing soda is associated with risk of hematopoetic cancers. We repeatedly assessed diet in the Nurses' Health Study (NHS) and
Marina Marinovich et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 60, 109-115 (2013-07-31)
Aspartame is a synthetic sweetener that has been used safely in food for more than 30 years. Its safety has been evaluated by various regulatory agencies in accordance with procedures internationally recognized, and decisions have been revised and updated regularly.
L D Stegink
The American journal of clinical nutrition, 46(1 Suppl), 204-215 (1987-07-01)
Toxicology is based on the premise that all compounds are toxic at some dose. Thus, it is not surprising that very large doses of aspartame (or its components--aspartate, phenylalanine, and methanol) produce deleterious effects in sensitive animal species. The critical
T J Maher et al.
Environmental health perspectives, 75, 53-57 (1987-11-01)
The artificial sweetener aspartame (L-aspartyl-L-phenylalanyl-methyl ester), is consumed, primarily in beverages, by a very large number of Americans, causing significant elevations in plasma and, probably, brain phenylalanine levels. Anecdotal reports suggest that some people suffer neurologic or behavioral reactions in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service