Skip to Content
Merck
All Photos(1)

Key Documents

45706

Supelco

Warfarin

Warfarin

PESTANAL®, analytical standard

Synonym(s):

4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin, Coumafene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H16O4
CAS Number:
Molecular Weight:
308.33
Beilstein:
8868198
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

162-164 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

CC(=O)CC(c1ccccc1)C2=C(O)c3ccccc3OC2=O

InChI

1S/C19H16O4/c1-12(20)11-15(13-7-3-2-4-8-13)17-18(21)14-9-5-6-10-16(14)23-19(17)22/h2-10,15,21H,11H2,1H3

InChI key

PJVWKTKQMONHTI-UHFFFAOYSA-N

Gene Information

human ... VKORC1(79001)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Warfarin is an anticoagulant commonly used in the prevention of thrombosis and thromboembolism. Its use is studied in stroke and pulmonary fibrosis patients among many others. It is used to study potential risks and interactions with other drugs and food[1].

Biochem/physiol Actions

Warfarin consists of a racemic mixture of two active enantiomers, R- and S- forms. Warfarin is slower-acting than the common anticoagulant heparin. Warfarin inhibits the vitamin K-dependent synthesis of biologically active forms of the calcium-dependent clotting factors II, VII, IX and X. It inhibits regulatory factors protein C, protein S, and protein Z[2].

Caution

Restricted to professional users. Attention − Avoid exposure − obtain special instructions before use.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - Repr. 1A - STOT RE 1 Oral

Target Organs

Blood

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Jack Ansell et al.
Chest, 126(3 Suppl), 204S-233S (2004-09-24)
This article concerning the pharmacokinetics and pharmacodynamics of vitamin K antagonists (VKAs) is part of the Seventh American College of Chest Physicians Conference on Antithrombotic and Thrombolytic Therapy: Evidence-Based Guidelines. The article describes the antithrombotic effect of VKAs, the monitoring
A Placebo-Controlled Randomized Trial of Warfarin in Idiopathic Pulmonary Fibrosis
Imre Noth, Kevin J. Anstrom, et al.
American Journal of Respiratory and Critical Care Medicine, 10.1164, 201202-03140C-201202-03140C (2012)
Periprocedural management of new oral anticoagulants in patients undergoing atrial fibrillation ablation.
Jeffrey I Weitz et al.
Circulation, 129(16), 1688-1694 (2014-04-23)
Daisuke Yoshioka et al.
Journal of artificial organs : the official journal of the Japanese Society for Artificial Organs, 17(4), 308-314 (2014-07-23)
The aim of this study was to evaluate our clinical experience with the Jarvik 2000 axial flow pump (Jarvik Heart, Inc, New York, NY, USA), a miniature axial flow left ventricular assist device (LVAD). The clinical results of eight patients
Aaron Liew et al.
The Canadian journal of cardiology, 29(7 Suppl), S34-S44 (2013-06-29)
Warfarin is effective for the prevention and treatment of thromboembolism but produces variable anticoagulant effects and requires routine monitoring of the international normalized ratio (INR) to optimize the balance between efficacy and safety. The new oral anticoagulants (NOACs) have a

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service