Skip to Content
Merck
All Photos(2)

Key Documents

U105

Sigma-Aldrich

U-62066

solid

Synonym(s):

(±)-(5α,7α,8β)-3,4-Dichloro-N-methyl-N-[7-(1-pyrrolidinyl)-1-oxaspiro[4.5]dec-8-yl]benzeneacetamide mesylate salt, Spiradoline mesylate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C22H30Cl2N2O2 · CH3SO3H
CAS Number:
Molecular Weight:
521.50
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

color

white

solubility

H2O: 14 mg/mL
methanol: insoluble

storage temp.

2-8°C

SMILES string

CS(O)(=O)=O.CN([C@H]1CC[C@@]2(CCCO2)C[C@@H]1N3CCCC3)C(=O)Cc4ccc(Cl)c(Cl)c4

InChI

1S/C22H30Cl2N2O2.CH4O3S/c1-25(21(27)14-16-5-6-17(23)18(24)13-16)19-7-9-22(8-4-12-28-22)15-20(19)26-10-2-3-11-26;1-5(2,3)4/h5-6,13,19-20H,2-4,7-12,14-15H2,1H3;1H3,(H,2,3,4)/t19-,20-,22-;/m0./s1

InChI key

FHEZDPDAYTVKKG-JLBKCEDKSA-N

Gene Information

human ... OPRK1(4986)

Biochem/physiol Actions

U-62066, also known as spiradoline, is a highly selective κ opioid receptor agonist. It exhibits analgesic and diuretic effects. Antitussive property of U-62066 is observed in rats.

Preparation Note

U-62066 is soluble in water at 14 mg/ml, but is insoluble in methanol.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diana T McCloskey et al.
American journal of physiology. Heart and circulatory physiology, 294(1), H205-H212 (2007-10-30)
Increased signaling by G(i)-coupled receptors has been implicated in dilated cardiomyopathy. To investigate the mechanisms, we used transgenic mice that develop dilated cardiomyopathy after conditional expression of a cardiac-targeted G(i)-coupled receptor (Ro1). Activation of G(i) signaling by the Ro1 agonist
Kimberly Scearce-Levie et al.
BMC biology, 3, 3-3 (2005-02-15)
The physiological regulation of G protein-coupled receptors, through desensitization and internalization, modulates the length of the receptor signal and may influence the development of tolerance and dependence in response to chronic drug treatment. To explore the importance of receptor regulation
M F Piercey et al.
The Journal of pharmacology and experimental therapeutics, 251(1), 267-271 (1989-10-01)
Administered i.p. to mice, the selective kappa receptor agonists U-50488H and spiradoline (U-62066) were more potent on the tail-flick than on the hot-plate analgesic assay. Both were more potent after i.s. rather than i.c. administration, a result consistent with earlier
U-47700 and Its Analogs: Non-Fentanyl Synthetic Opioids Impacting the Recreational Drug Market.
Baumann, et al.
Brain sciences, 10 (2020)
Daniela Braida et al.
British journal of pharmacology, 157(5), 844-853 (2009-05-09)
Drugs targeting brain kappa-opioid receptors produce profound alterations in mood. In the present study we investigated the possible anxiolytic- and antidepressant-like effects of the kappa-opioid receptor agonist salvinorin A, the main active ingredient of Salvia divinorum, in rats and mice.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service