Skip to Content
Merck

Skip To

OLIGO

Custom DNA Oligos

Design and Order Custom Oligos

Synonym(s):

Custom DNA Oligos, DNA, DNA primer, High-throughput DNA, ISO 13485 Oligos, Locked Nucleic Acid oligos, Modified oligos, NGS adapters, NGS−grade oligos, Next−generation sequencing adapters, Next−generation sequencing oligos, Oligonucleotide, Oligonucleotides, Oligos, Oligos in plates, cGMP Oligos

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
41105327


      


Standard DNA Oligos

  • Scale: 0.025, 0.05, 0.2, 1.0, 10, and 15 μmol
  • Purification: Desalt, Cartridge, HPLC, and PAGE
  • Sequence Length: Up to 120 bases
  • Modifications: Over 200 available, including dyes (fluorescent and non-fluorescent)
  • Quality Control: 100% Mass Spectrometry
  • Format: Supplied dry or in solution – tubes, plates and mixed plates (forward and reverse primers)
  • WellRED Oligos: Available
  • qPCR Probes: Available
  • Learn more about Standard DNA Synthesis

Tubes

Plates


Easy Oligo

  • Amount: 3 OD
  • Concentration: 100 µM
  • Purification: Desalt
  • Sequence Length: 15 - 30 bases
  • Modifications: Not available
  • Quality Control: 100% Mass Spectrometry
  • Format: Supplied in solution
  • Ships within 24 hours of order receipt
  • Learn more

Tubes


Pure & Simple Primers

  • Amount: 3 OD
  • Purification: Cartridge
  • Sequence Length: 12 - 35 bases
  • Modifications: Not available
  • Quality Control: 100% Mass Spectrometry
  • Format: Supplied dry
  • Ships within 24 hours of order receipt
  • Learn more

Tubes


iScale Oligos™ (mg & g quantity)

  • Larger quantities for in vivo, high throughput and commercial projects
  • Quantity: 10, 25, 50, 100, 250, 500 mg; 1, 2, 5, 10, 15 g
  • Purification: Desalt, RP-HPLC & In vivo treatment
  • Sequence Length: 10 - 55 bases (Inquire for longer lengths)
  • Modifications: 6-FAM, Biotin, Phosphate, Amino-Modifiers & others
  • Quality Control: 100% Mass Spectrometry
  • Format: Supplied dry
  • Services: Salt Exchange, Filtration & Endotoxin Testing
  • Learn more

Tubes


Next-Gen Sequencing Oligos

  • Custom universal and index (barcode) adapter oligos for major sequencing platforms
  • Suitable purity (percent full-length sequence) to maintain sequence read integrity
  • Low levels of cross-contamination to facilitate proper sequence assembly
  • Learn more

Commercial Oligos

  • Custom DNA Primers, qPCR Probes, and Next-Gen Sequencing Oligos for Life Science Research Tools, Molecular Diagnostics (MDx), and Laboratory Developed Tests (LDTs)
  • Component or complete kit manufacturing, including custom formulations, packaging, and private labeling
  • Learn more

GMP OLIGOS

  • Custom cGMP Oligos, available with custom specifications, are Commercial Oligos (raw materials) for MDx and LDTs
  • Learn more

Locked Nucleic Acid

  • Available with Custom DNA Oligos
  • Increased thermal stability and hybridization specificity
  • Easier and more specific designs for problematic target sequences
  • Learn more

Tubes

Sign In To Continue

To continue reading please sign in or create an account.

Don't Have An Account?

Fuqiang Chen et al.
Nature methods, 8(9), 753-755 (2011-07-19)
Zinc-finger nucleases (ZFNs) have enabled highly efficient gene targeting in multiple cell types and organisms. Here we describe methods for using simple ssDNA oligonucleotides in tandem with ZFNs to efficiently produce human cell lines with three distinct genetic outcomes: (i)
T N Wiegner et al.
Marine pollution bulletin, 114(2), 952-961 (2016-11-22)
Spatial and temporal patterns of coastal microbial pollution are not well documented. Our study examined these patterns through measurements of fecal indicator bacteria (FIB), nutrients, and physiochemical parameters in Hilo Bay, Hawai'i, during high and low river flow. >40% of
Leonel Armas-López et al.
Oncotarget, 8(40), 67056-67081 (2017-10-06)
Several homeobox-related gene (HOX) transcription factors such as mesenchyme HOX-2 (MEOX2) have previously been associated with cancer drug resistance, malignant progression and/or clinical prognostic responses in lung cancer patients; however, the mechanisms involved in these responses have yet to be



Global Trade Item Number

SKUGTIN
O9140-5MG04061834248811
69896-304055977258776
69593-304055977274462
LMO001-30RXN04065267221786
LMO001-100RXN04065267221779
930482-1G04065268833575
O4387-.1ML04061834233831
S720004053252725159
932574-1G04065269291848
GE3815210301035004061837623967
80803404061833671368
GE3815210301115004061837623974
GE3815210301015004061837623950
80803104061833671306
70030-U04065272632386
70029-U04065272632379
70028-U04065272632362
70000-U04065272631884
70001-U04065272631891
70011-U04065272632195
70007-U04065272632157
70010-U04065272632188
70002-U04065272631907
70006-U04065272632140
70004-U04065272632126