Skip to Content
Merck
All Photos(4)

Key Documents

F6886

Sigma-Aldrich

Forskolin

from Coleus forskohlii, ≥98% (HPLC), powder, positive ionotropic agent

Synonym(s):

7β-Acetoxy-8,13-epoxy-1α,6β,9α-trihydroxylabd-14-en-11-one, Coleonol, Colforsin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H34O7
CAS Number:
Molecular Weight:
410.50
Beilstein:
1692716
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

Product Name

Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder

biological source

Coleus forskohlii

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

ethanol: soluble 50 mg/mL

SMILES string

CC(=O)O[C@H]1[C@@H](O)[C@H]2C(C)(C)CC[C@H](O)[C@]2(C)[C@@]3(O)C(=O)C[C@@](C)(O[C@]13C)C=C

InChI

1S/C22H34O7/c1-8-19(5)11-14(25)22(27)20(6)13(24)9-10-18(3,4)16(20)15(26)17(28-12(2)23)21(22,7)29-19/h8,13,15-17,24,26-27H,1,9-11H2,2-7H3/t13-,15-,16-,17-,19-,20-,21+,22-/m0/s1

InChI key

OHCQJHSOBUTRHG-KGGHGJDLSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Forskolin, present predominantly in the root of Coleus forskohlii plant, is a potential pharmacological compound. It is used for treating hypertension, respiratory disorders and obesity. Forskolin is a diterpenoid capable of modulating adenylate cyclase enzyme and cyclic adenosine monophosphate (cAMP) levels. Engineering forskolin biosynthetic pathway caters its commercial production for pharmacological use. Forskolin has antioxidant and anti-inflammatory property and reduces hyperglycemia by stimulating insulin release. Forskolin promotes differentiation of hepatoma HepaRG cells and activates farnesoid X receptor (FXR) and pregnane X receptor(PXR).
Several studies indicate that forskolin is linked to increased lipolysis, potentially leading to increased fat and weight reduction within the body. The supplementation of forskolin is believed to potentially improve fat loss without losing muscle mass.

Application

Forskolin has been used:
  • for inducing mRNA expression in primary hepatocytes.
  • as a defatting drug in primary human hepatocytes.
  • as a medium supplement to induce differentiation of adipose-derived stem cells (ASCs).
  • in the activation of the cystic fibrosis transmembrane conductance regulator (CFTR) gene.
  • to promote the differentiation of human induced pluripotent stem cells (hiPSCs) into endothelial cells.
  • to stimulate cystic fibrosis transmembrane conductance regulator (CFTR)-mediated chloride ion secretion in differentiated airway epithelial cells derived from individuals with and without cystic fibrosis (CF).

Biochem/physiol Actions

Cell-permeable diterpenoid that possesses anti-hypertensive, positive inotropic, and adenylyl cyclase activating properties. Many of its biological effects are due to its activation of adenylyl cyclase and the resulting increase in intracellular cAMP concentration. Forskolin effects calcium currents and inhibits MAP kinase.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Adenylyl cyclases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

Forskolin is soluble in organic solvents such as ethanol, chloroform, and DMSO.

Storage and Stability

Forskolin solutions at 5 mg/mL in either ethanol or DMSO are stable for at least 6 months at room temperature per HPLC analysis.
This product is to be stored in room temperature . Store the product tightly closed under dry condition.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functional polarization of human hepatoma HepaRG cells in response to forskolin
Mayati A, et al.
Scientific Reports, 8(1), 16115-16115 (2018)
Use of a high-throughput phenotypic screening strategy to identify amplifiers, a novel pharmacological class of small molecules that exhibit functional synergy with potentiators and correctors
Giuliano KA, et al.
SLAS discovery, 23(2), 111-121 (2018)
Effect of chronic administration of forskolin on glycemia and oxidative stress in rats with and without experimental diabetes
Rios-Silva M, et al.
International Journal of Medical Sciences, 11(5), 448-448 (2014)
Total biosynthesis of the cyclic AMP booster forskolin from Coleus forskohlii
Pateraki I, et al.
eLife, 6, e23001-e23001 (2017)
Differentiation of adipose-derived stem cells into Schwann cell-like cells through intermittent induction: potential advantage of cellular transient memory function
Sun X, et al.
Stem Cell Research & Therapy, 9(1), 133-133 (2018)

Articles

Naive pluripotent stem cells cultured in vitro using specialized media and inhibitors mimic "ground-state" cells from blastocysts.

Naive pluripotent stem cells cultured in vitro using specialized media and inhibitors mimic "ground-state" cells from blastocysts.

Naive pluripotent stem cells cultured in vitro using specialized media and inhibitors mimic "ground-state" cells from blastocysts.

Naive pluripotent stem cells cultured in vitro using specialized media and inhibitors mimic "ground-state" cells from blastocysts.

See All

Protocols

A rapid in vitro assay for CFTR function, the forskolin-induced swelling protocol uses human colon organoids, which can be derived from cystic fibrosis patient tissue.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service