Skip to Content
Merck

42874

Tetramethylrhodamine isothiocyanate–Dextran

average mol wt 40,000

Synonym(s):

TRITC-Dextran 40, TRITC-Dextran

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352116
NACRES:
NA.32
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Segment

form

powder

mol wt

average mol wt 40,000

extent of labeling

0.001-0.008 mol/mol TRITC glucose

fluorescence

λex 550 nm; λem 577 nm±5 nm in 0.1 M Tris pH 8.0

application(s)

cell analysis

General description

Tetramethylrhodamine isothiocyanate–Dextran, also known as TRITC-Dextran or TRITC-Dextran 40, is a fluorescent dye and a novel drug. The fluorescent activity of TRITC-Dextran is detected at 555nm wavelength. The TRITC-Dextran molecules are released in the cells through passive diffusion due to a concentration gradient.

Application

Tetramethylrhodamine isothiocyanate–Dextran (TRITC-Dextran 40) is used as a fluorescent probe to study cell processes such as cell permeability, phagocytosis and endocytosis and to study mechanisms of biomolecular drug delivery.


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library