Skip to Content
Merck
All Photos(2)

Documents

D2926

Sigma-Aldrich

Diphenyleneiodonium chloride

≥98%

Synonym(s):

DPI, Dibenziodolium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8ClI
CAS Number:
Molecular Weight:
314.55
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98%

form

powder

mp

278 °C

solubility

DMSO: 9.80-10.20 mg/mL, clear, colorless to faintly yellow

storage temp.

room temp

SMILES string

[Cl-].[I+]1c2ccccc2-c3ccccc13

InChI

1S/C12H8I.ClH/c1-3-7-11-9(5-1)10-6-2-4-8-12(10)13-11;/h1-8H;1H/q+1;/p-1

InChI key

FCFZKAVCDNTYID-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Diphenyleneiodonium chloride has been used as an NOX inhibitor:
  • to study its effects on the reactive oxygen species (ROS) levels and also its inhibitory effects on killing tumor cells by neutrophils isolated from clinical malignant pleural effusion (MPE) samples
  • as a nicotinamide adenine dinucleotide phosphate (NADPH) oxidase inhibitor to confirm the variation in inflammation was secondary to overproduction of reactive oxygen species (ROS) in cells treated with doxorubicin (DOX) + flagellin (FL)
  • as an NADPH oxidase (NOX) inhibitor to study its effects on palmitate (PA)-induced sphere-forming ability of HepG2 (hepatocellular carcinoma) cells

Biochem/physiol Actions

A potent and reversible inhibitor of nitric oxide synthetase from macrophages and endothelial cells. Also inhibits other flavoenzymes such as neutrophil NADPH oxidase.
Diphenyleneiodonium (DPI) can affect mitochondrial function. DPI can block neuron degeneration in Parkinson′s disease. It provides neuroprotection following focal cerebral ischemia. DPI can induce a chemo-quiescence phenotype in cancer stem-like cells, effectively inhibiting their proliferation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ching-Hou Ma et al.
Redox biology, 14, 72-81 (2017-09-05)
Osteoarthritis (OA) is a degenerative joint disease affecting a large population of people. Although the elevated expression of PKR (double stranded RNA-dependent protein kinase) and MMP-13 (collagenase-3) have been indicated to play pivotal roles in the pathogenesis of OA, the
Mito Kanatsu-Shinohara et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(33), 16404-16409 (2019-07-31)
Because spermatogonial stem cells (SSCs) are immortal by serial transplantation, SSC aging in intact testes is considered to be caused by a deteriorated microenvironment. Here, we report a cell-intrinsic mode of SSC aging by glycolysis activation. Using cultured SSCs, we
Guei-Sheung Liu et al.
Journal of tissue engineering and regenerative medicine, 10(3), E167-E176 (2013-07-31)
Adipose-derived stem cells (ASCs) show great potentials in applications such as therapeutic angiogenesis, regenerative medicine and tissue engineering. Pharmacological preconditioning of stem cells to boost the release of cytoprotective factors may represent an effective way to enhance their therapeutic efficacy.
Clara Angelina et al.
Scientific reports, 7(1), 16867-16867 (2017-12-06)
Relapse-prone, poor prognosis neuroblastoma is frequently characterized by deletion of chr1p36 where tumor suppressor gene KIF1Bβ resides. Interestingly, many 1p36-positive patients failed to express KIF1Bβ protein. Since altered cellular redox status has been reported to be involved in cell death
Lingdi Wang et al.
Nature communications, 8(1), 523-523 (2017-09-14)
The mitochondrial enriched GCN5-like 1 (GCN5L1) protein has been shown to modulate mitochondrial protein acetylation, mitochondrial content and mitochondrial retrograde signaling. Here we show that hepatic GCN5L1 ablation reduces fasting glucose levels and blunts hepatic gluconeogenesis without affecting systemic glucose

Articles

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service