Skip to Content
Merck

Skip To

T7765

Tunicamycin

from Streptomyces sp., ≥98% (HPLC), powder, N-acetylglucosamine transferase inhibitor

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77
Beilstein/REAXYS Number:
6888090
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Tunicamycin from Streptomyces sp.,

Quality Segment

solubility

95% ethanol: soluble 1 mg/mL, clear to faintly hazy, THF: soluble <1 mg/mL, dioxane: soluble <1 mg/mL, DMF: soluble >10 mg/mL, pyridine: >10 mg/mL, DMSO: soluble 4.9-5.1 mg/mL, clear to slightly hazy, colorless to yellow, methanol: slightly soluble 4.9-5.1 mg/mL, methanol: soluble 4.9-5.1 mg/mL, clear to slightly hazy, colorless to yellow, acetone: insoluble, aqueous base: insoluble, chloroform: insoluble, ethyl acetate: insoluble

antibiotic activity spectrum

fungi, viruses

mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

CC(C)CCCCCCCC\C=C\C(=O)N[C@@H]1[C@@H](O)[C@@H](O)[C@@H](C[C@@H](O)[C@H]2O[C@H]([C@H](O)[C@@H]2O)N3C=CC(=O)NC3=O)O[C@H]1O[C@@H]4O[C@@H](CO)[C@H](O)[C@@H](O)[C@@H]4NC(C)=O

InChI

1S/C37H60N4O16/c1-18(2)12-10-8-6-4-5-7-9-11-13-23(45)39-26-30(50)27(47)21(54-36(26)57-35-25(38-19(3)43)29(49)28(48)22(17-42)55-35)16-20(44)33-31(51)32(52)34(56-33)41-15-14-24(46)40-37(41)53/h11,13-15,18,20-22,25-36,42,44,47-52H,4-10,12,16-17H2,1-3H3,(H,38,43)(H,39,45)(H,40,46,53)/b13-11+/t20-,21-,22+,25+,26-,27+,28+,29+,30-,31+,32-,33-,34-,35+,36+/m1/s1

InChI key

YJQCOFNZVFGCAF-WPTOCQRYSA-N

General description

Chemical structure: nucleoside
Contains homologues A,B,C, and D. Composition may vary from lot to lot. Actual content given on label.
Tunicamycin is an antibiotic and a glycosylation inhibitor. Its mode of action includes blocking human UDP-HexNAc:polyprenol-P HexNAc-1-P enzymes in the enzyme GlcNAc phosphotransferase (GPT). Glycosylation is a major post-translational modification that plays a critical role in glycoprotein folding, stability, and subcellular localization, as well as in the biological functions of the glycoprotein. Abnormal glycosylation has been identified as a signature of cancer and has been shown to play a significant role in tumor progression, metastasis, and chemoresistance. Research shows tunicamycin exhibits antitumor action. Furthermore, tunicamycin inhibition of N-glycosylation ultimately results in the build-up of unstructured proteins in the endoplasmic reticulum (ER) lumen, potentially leading to ER stress.[1] Tunicamycin also exhibits antibacterial and antifungal actions. It blocks the formation of protein N-glycosidic linkages by inhibiting the transfer of N-acetylglucosamine 1-phosphate to dolichol monophosphate. Tunicamycin also Inhibits bacterial and eukaryote N-acetylglucosamine transferases and prevents the formation of N-acetylglucosamine lipid intermediates.

Application

Tunicamycin has been used to study the effect of N-linked glycosylation of human proton-coupled folate transporter (HsPCFT) in HeLa cells[2]. Tunicamycin has also been used to study the functional effects of coxsackievirus and adenovirus receptor (CAR) glycosylation in COS-7 cells[3]. Tunicamycin from Streptomyces sp. has been used to inhibit N-glycosylation in cell culture experiments.

Biochem/physiol Actions

Blocks the formation of protein N-glycosidic linkages.

Preparation Note

Tunicamycin dissolves in DMSO at 4.9-5.1 mg/ml and yields a clear to very slightly hazy, colorless to yellow solution. Furthermore, tunicamycin is soluble in DMF (>10 mg/ml), pyridine (>10 mg/ml), water (<5 mg/ml, pH 9.0), dioxane (<1 mg/ml) and THF. However, it is insoluble in other organic solvents such as acetone, chloroform, and ethyl acetate, and in aqueous solutions with pH <6. Aqueous solutions can be prepared from stock solutions by diluting with water at pH 8-10 or with buffers with pH >7, preferably >8. Tunicamycin will not dissolve in phosphate buffer, pH 8, at 1 mg/ml, even with heating, but solubility can be achieved by raising the pH to 9 and back titrating to pH 7-8.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
654380T4540647925
description

-

description

-

description

-

description

A potent and reversible, cell-permeable inhibitor of histone deacetylase.

Quality Level

300

Quality Level

200

Quality Level

200

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

solubility

95% ethanol: soluble 1 mg/mL, clear to faintly hazy, dioxane: soluble <1 mg/mL, pyridine: >10 mg/mL, methanol: slightly soluble 4.9-5.1 mg/mL, acetone: insoluble, chloroform: insoluble, THF: soluble <1 mg/mL, DMSO: soluble 4.9-5.1 mg/mL, clear to slightly hazy, colorless to yellow, aqueous base: insoluble, DMF: soluble >10 mg/mL, ethyl acetate: insoluble, methanol: soluble 4.9-5.1 mg/mL, clear to slightly hazy, colorless to yellow

solubility

DMSO: 10 mg/mL, DMF: soluble, pyridine: soluble

solubility

methanol: soluble 4.90-5.10 mg/mL, clear (Pale yellow to yellow), methanol: soluble 4.90-5.10 mg/mL, clear, pale yellow to yellow

solubility

ethanol: 1 mg/mL, DMSO: 20 mg/mL, methanol: soluble

mode of action

protein synthesis | interferes

mode of action

-

mode of action

enzyme | inhibits

mode of action

-

antibiotic activity spectrum

fungi

antibiotic activity spectrum

-

antibiotic activity spectrum

-

antibiotic activity spectrum

-


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 1 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. Is it possible to store the product at -20/30°C for extended storage?

    1 answer
    1. Store this product at 2–8 °C. When stored properly, the product remains active for at least 3 years. Solutions of tunicamycin in DMSO (5 mg/ml) can be stored at -20 °C in small aliquots of 25 - 50uL. No specific time frame for storage at -20 °C was indicated.

      Helpful?

  2. What is the molecular weight for T7765 product ?

    1 answer
    1. The molecular weight of the product is lot-specific. Each homolog has a specific molecular weight: A = 817, B = 831, C = 845, and D = 859. The lot-specific Certificate of Analysis (COA) provides the percentage contribution for each homolog.

      Helpful?

Reviews

No rating value

Active Filters