Skip to Content
Merck
All Photos(1)

Documents

SML0064

Sigma-Aldrich

MBX-102

≥98% (HPLC)

Synonym(s):

(aR)-4-Chloro-a-[3-(trifluoromethyl)phenoxy]benzeneacetic acid 2-(acetylamino)ethyl ester, Arhalofenate, JNJ39659100, MBX-102/JNJ39659100

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17ClF3NO4
CAS Number:
Molecular Weight:
415.79
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥35 mg/mL

originator

Johnson & Johnson

storage temp.

2-8°C

InChI

1S/C19H17ClF3NO4/c1-12(25)24-9-10-27-18(26)17(13-5-7-15(20)8-6-13)28-16-4-2-3-14(11-16)19(21,22)23/h2-8,11,17H,9-10H2,1H3,(H,24,25)/t17-/m1/s1

InChI key

BJBCSGQLZQGGIQ-QGZVFWFLSA-N

Application

MBX-102 may be used in PPAR-mediated cell signaling studies.

Biochem/physiol Actions

MBX-102 has a potent transrepression effect on PPARγ. It is an oral glucose-reducing agent and also has insulin-sensitizing properties. It is useful as treatment for type 2 diabetes. MBX-102 also lowers triglycerides in a PPARα-independent manner.
MBX-102 is a selective PPAR modulator (SPPARM) and has been shown to inhibit phosphorylation of PPARγ. MBX-102 is converted to the active form, MBX-102 acid, in vivo.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Nuclear Receptors (Non-Steroids) and Nuclear Receptors (PPARs) pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Apurva Chandalia et al.
PPAR research, 2009, 706852-706852 (2009-05-01)
MBX-102/JNJ-39659100 (MBX-102) is a selective, partial PPAR-γ agonist that lowers glucose in the absence of some of the side effects, such as weight gain and edema, that are observed with the TZDs. Interestingly MBX-102 also displays pronounced triglyceride lowering in
Francine M Gregoire et al.
Molecular endocrinology (Baltimore, Md.), 23(7), 975-988 (2009-04-25)
MBX-102/JNJ39659100 (MBX-102) is in clinical development as an oral glucose-lowering agent for the treatment of type 2 diabetes. MBX-102 is a nonthiazolidinedione (TZD) selective partial agonist of peroxisome proliferator-activated receptor (PPAR)-gamma that is differentiated from the TZDs structurally, mechanistically, preclinically

Articles

Peroxisome proliferator activated receptors (PPARs) are ligand-activated transcription factors related to hormone receptors, influencing gene expression.

Peroxisome proliferator activated receptors (PPARs) are ligand-activated transcription factors related to hormone receptors, influencing gene expression.

Peroxisome proliferator activated receptors (PPARs) are ligand-activated transcription factors related to hormone receptors, influencing gene expression.

Peroxisome proliferator activated receptors (PPARs) are ligand-activated transcription factors related to hormone receptors, influencing gene expression.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service