491578
4-Methoxybenzoic acid-α-13C
99 atom % 13C
Synonym(s):
p-Anisic acid-carboxy-13C
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3O-C6H4-13CO2H
CAS Number:
Molecular Weight:
153.14
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
Recommended Products
isotopic purity
99 atom % 13C
Quality Level
Assay
99% (CP)
form
solid
mp
183-184 °C (lit.)
mass shift
M+1
SMILES string
COc1ccc(cc1)[13C](O)=O
InChI
1S/C8H8O3/c1-11-7-4-2-6(3-5-7)8(9)10/h2-5H,1H3,(H,9,10)/i8+1
InChI key
ZEYHEAKUIGZSGI-VJJZLTLGSA-N
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
365.0 °F - closed cup
Flash Point(C)
185.00 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service