Skip to Content
Merck
All Photos(2)

Documents

SAB4503329

Sigma-Aldrich

Anti-DARPP-32 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Dopamine- and cAMP-regulated neuronal phosphoprotein, IPPD, Neuronal phosphoprotein DARPP-32, PPP1R1B

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 22 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PPP1R1B(84152)

General description

Anti-DARPP-32 Antibody detects endogenous levels of total DARPP-32 protein.
PPP1R1B (protein phosphatase 1 regulatory inhibitor subunit 1B)/DARPP32 (dopamine- and cAMP-regulated phosphoprotein of molecular weight 32 kDa) encodes two transcripts which are further translated into two DARPP-32 protein isoforms, full length (fl)-DARPP-32 and truncated (t)-DARPP. It is located on human chromosome 17q12.
The DARPP-32 (dopamine and cAMP-regulated neuronal phosphoprotein 2) gene is mapped to human chromosome 17q12. The gene is expressed predominantly in spiny neurons of the neostriatum and specifically localized in caudate, putamen, nucleus accumbens, cerebral and cerebellar cortex. The gene encodes two isoforms, full length (fl)-DARPP-32 and truncated (t)-DARPP. The lack of the first 36 residues at the N-terminal in t- DARPP marks the difference between the two isoforms. Anti-DARPP-32 Antibody detects endogenous levels of total DARPP-32 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human DARPP-32.

Immunogen Range: 41-90

Biochem/physiol Actions

DARPP-32 (dopamine and cAMP-regulated neuronal phosphoprotein 2) is a dopaminergic gene that influences dopamine receptor binding potential in the corpus striatum. The gene also regulates the availability of dopamine neurotransmitter, which is important for cognitive capacities. Therefore, variation in this gene affects category learning performance. (t)-DARPP takes part in the process of tumorigenesis by stimulating chemokine receptor 4 and also promoting cell proliferation. Elevated expression of the DARPP-32 gene in dorsolateral prefrontal neocortex and hippocampus is correlated with the development of schizophrenia and cognitive disorders. The encoded protein is a phosphorylation-dependent inhibitor. DARPP-32 is associated with the pathogenesis of gastric cancer and breast cancer with its anti-apoptotic property, and is found to promote angiogenesis.
PPP1R1B (protein phosphatase 1 regulatory inhibitor subunit 1B)/DARPP32 (dopamine- and cAMP-regulated phosphoprotein of molecular weight 32 kDa) is considered as an inhibitor of phosphate-1 (PP-1) protein. It is associated with working memory and emotion processing.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jay V Deng et al.
Neuroscience, 369, 212-221 (2017-11-21)
Many neuropsychiatric disorders are associated with abnormal decision making involving risk of punishment, but the underlying molecular basis remains poorly understood. Methyl CpG-binding protein 2 (MeCP2) is an epigenetic factor that regulates transcription by directly binding to methylated DNA. Here
Gastric tumour-derived ANGPT2 regulation by DARPP-32 promotes angiogenesis.
Chen Z, et al.
Gut (2015)
Darpp-32: a novel antiapoptotic gene in upper gastrointestinal carcinomas.
Belkhiri A, et al.
Cancer Research, 65(15), 6583-6592 (2005)
Integrated gene copy number and expression microarray analysis of gastric cancer highlights potential target genes.
Myllykangas S, et al.
International Journal of Cancer. Journal International Du Cancer, 123(4), 817-825 (2008)
Dopaminergic genetic polymorphisms predict rule-based category learning.
Byrne K A, et al.
Journal of Cognitive Neuroscience, 28(7), 959-970 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service