Skip to Content
Merck
All Photos(3)

Documents

AB10518

Sigma-Aldrich

Anti-DARPP-32 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

protein phosphatase 1, regulatory (inhibitor) subunit 1B, dopamine and cAMP-regulated neuronal phosphoprotein 32, dopamine and cAMP regulated phosphoprotein, protein phosphatase 1 regulatory subunit 1B, Dopamine- and cAMP-regulated neuronal phosphoprotei

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse, rhesus macaque

species reactivity (predicted by homology)

canine (based on 100% sequence homology), rhesus monkey (based on 100% sequence homology)

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Ppp1R1B(19049)

General description

Dopamine- and cAMP-regulated neuronal phosphoprotein (DARP-32) is a cytoplasmic member of the protein phosphatase inhibitor 1 family. DARP-32 is a multifunctional protein kinase and protein phosphatase regulator, depending on the site of its activation. It possesses at least four sites that can be phosphorylated. Phosphorylation by PKA at Thr34 stimulates DARP-32 to act as a protein-phosphatase-1 (PP-1) inhibitor. However, phosphorylation at Thr75 will inhibit Thr34 phosphorylation. This selective, bidirectional regulation allows DARPP-32 to have a wide range of influence on downstream physiological effectors such as: ion pumps, neurotransmitter receptors, voltage-gate ion channels, and transcriptional factors. Studies suggest that it may also have a role in the Circadian System photic pathway. Overexpression of DARPP-32 has been linked with antiapoptotic mechanisms of cancer cells and has been observed in breast, colon, esophagus, gastric, and prostate cancer tissues.

Specificity

This antibody recognizes DARPP-32 near the N-terminus.

Immunogen

KLH-conjugated linear peptide corresponding to mouse DARPP-32 near the N-terminus.

Application

Anti-DARPP-32 Antibody is an antibody against DARPP-32 for use in WB & IP.
Research Sub Category
Neurotransmitters & Receptors

Ion Channels & Transporters
Western Blot (Snap i.d.) Analysis: 1 µg/mL of this antibody from a previous lot detected DARPP-32 on 10 µg of rat brain tissue lysate.

Immunoprecipitaion Analysis: 10 µg of this antibody from a previous lot immunoprecipitated DARPP-32 from rat brain RIPA lysate.

Quality

Evaluated by Western Blot on rat brain tissue lysate.

Western Blot Analysis: 0.1 µg/mL of this antibody detected DARPP-32 on 10 µg of rat brain tissue lysate.

Target description

~ 32 kDa observed

Linkage

Replaces: AB1656

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Despina E Ganella et al.
Frontiers in behavioral neuroscience, 11, 76-76 (2017-05-26)
Anxiety disorders are the most common type of mental disorder during adolescence, which is at least partly due to the resistance to extinction exhibited at this age. The dopaminergic system is known to be dysregulated during adolescence; therefore, we aimed
Inês T Afonso et al.
International journal of molecular sciences, 23(19) (2022-10-15)
Spinocerebellar ataxia type 2 (SCA2) is a rare autosomal, dominantly inherited disease, in which the affected individuals have a disease onset around their third life decade. The molecular mechanisms underlying SCA2 are not yet completely understood, for which we hypothesize
Stefanie L O'Neal et al.
Neurotoxicology, 44, 303-313 (2014-08-15)
Manganese (Mn) is an essential trace element, but excess exposure leads to accumulation in biological tissues, including the brain. Chronically high Mn levels in the brain are neurotoxic and can result in a progressive, irreversible neurological disorder known as manganism.
Daniel Enterría-Morales et al.
PloS one, 11(10), e0164391-e0164391 (2016-10-16)
Gender difference in Parkinson's disease (PD) suggests that female sex steroids may promote dopaminergic neuron survival and protect them from degeneration. The glial cell line-derived neurotrophic factor (GDNF) is believed to be dopaminotrophic; thus it is considered as a potential
Yukio Imamura et al.
Communications biology, 5(1), 636-636 (2022-06-30)
Synucleinopathies are neurodegenerative disorders including Parkinson disease (PD), dementia with Lewy body (DLB), and multiple system atrophy (MSA) that involve deposits of the protein alpha-synuclein (α-syn) in the brain. The inoculation of α-syn aggregates derived from synucleinopathy or preformed fibrils

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service