Skip to Content
Merck
All Photos(1)

Key Documents

S3255

Sigma-Aldrich

Sterigmatocystin

powder, ≥98% (TLC)

Synonym(s):

3a,12c-Dihydro-8-hydroxy-6-methoxy-7H-furol[3′,2′:4,5]furo[2,3-c]xanthen-7-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12O6
CAS Number:
Molecular Weight:
324.28
Beilstein:
53259
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (TLC)

form

powder

color

yellow

storage temp.

−20°C

SMILES string

COc1cc2OC3OC=CC3c2c4Oc5cccc(O)c5C(=O)c14

InChI

1S/C18H12O6/c1-21-11-7-12-13(8-5-6-22-18(8)24-12)17-15(11)16(20)14-9(19)3-2-4-10(14)23-17/h2-8,18-19H,1H3

InChI key

UTSVPXMQSFGQTM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Initiates lung and liver tumors under experimental conditions.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Motoyuki Shimizu et al.
Molecular and cellular biology, 32(18), 3743-3755 (2012-07-18)
Cellular levels of NAD(+) and NADH are thought to be controlled by de novo and salvage mechanisms, although evidence has not yet indicated that they are regulated by NAD(+) degradation. Here we show that the conserved nudix hydrolase isozyme NdxA
Christian Rank et al.
Fungal biology, 115(4-5), 406-420 (2011-05-03)
During the last 50y, the carcinogenic mycotoxin sterigmatocystin (ST) has been reported in several phylogenetically and phenotypically different genera: Aschersonia, Aspergillus, Bipolaris, Botryotrichum, Chaetomium, Emericella, Eurotium, Farrowia, Fusarium, Humicola, Moelleriella, Monocillium and Podospora. We have reexamined all available strains of
C N Ezekiel et al.
International journal of food microbiology, 162(3), 231-236 (2013-03-05)
In order to determine whether dried mushrooms are a foodstuff that may be less susceptible to infection by toxigenic molds and consequently to mycotoxin contamination, 34 dried market samples were analyzed. Fungal population was determined in the samples by conventional
Vellaisamy Ramamoorthy et al.
Molecular microbiology, 85(4), 795-814 (2012-07-13)
In Aspergillus nidulans the global regulatory gene veA is necessary for the biosynthesis of several secondary metabolites, including the mycotoxin sterigmatocystin (ST). In order to identify additional veA-dependent genetic elements involved in regulating ST production, we performed a mutagenesis on
Aleksandrs Versilovskis et al.
Molecular nutrition & food research, 54(1), 136-147 (2009-12-10)
Sterigmatocystin (STC) is a mycotoxin produced by fungi of many different Aspergillus species. Other species such as Bipolaris, Chaetomium, Emiricella are also able to produce STC. STC producing fungi were frequently isolated from different foodstuffs, while STC was regularly detected

Articles

Cancer research has revealed that the classical model of carcinogenesis, a three step process consisting of initiation, promotion, and progression, is not complete.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service