Skip to Content
Merck

Skip To

P5655

Potassium phosphate monobasic

powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%

Synonym(s):

Potassium dihydrogen phosphate, prim.-Potassium phosphate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
KH2PO4
CAS Number:
Molecular Weight:
136.09
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12161700
EC Number:
231-913-4
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99.0%

form

powder

technique(s)

cell culture | insect: suitable, cell culture | mammalian: suitable, cell culture | plant: suitable

pH

4.2-4.6 (20 °C, 20 g/L)

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

mp

252.6 °C (lit.)

solubility

water: 100 mg/mL, clear, colorless

density

2.338 g/mL at 25 °C (lit.)

application(s)

diagnostic assay manufacturing

storage temp.

room temp

SMILES string

[K+].OP(O)([O-])=O

InChI

1S/K.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

InChI key

GNSKLFRGEWLPPA-UHFFFAOYSA-M

Application

  • Potassium phosphate monobasic has been used during isolation of protein from bacterial cells.
  • It has been used in the preparation of PZM-3 (defined medium for porcine embryos) medium.[1]
  • It has been used in the preparation of phosphate buffer saline.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P9791RDD03704243
technique(s)

cell culture | insect: suitable, cell culture | plant: suitable, cell culture | mammalian: suitable

technique(s)

-

technique(s)

cell culture | insect: suitable, cell culture | mammalian: suitable, cell culture | plant: suitable

technique(s)

-

assay

≥99.0%

assay

≥98.0%

assay

≥99.0%

assay

98-100.5% (calc. to the dried substance)

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

200

application(s)

diagnostic assay manufacturing

application(s)

diagnostic assay manufacturing

application(s)

-

application(s)

-

form

powder

form

powder

form

-

form

crystalline

storage temp.

room temp

storage temp.

-

storage temp.

-

storage temp.

-


Still not finding the right product?

Explore all of our products under Potassium phosphate monobasic


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters