Skip to Content
Merck
All Photos(1)

Key Documents

32986

Supelco

Sterigmatocystin solution

~50 μg/mL in acetonitrile, analytical standard

Synonym(s):

3a,12c-Dihydro-8-hydroxy-6-methoxy-7H-furol[3′,2′:4,5]furo[2,3-c]xanthen-7-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12O6
CAS Number:
Molecular Weight:
324.28
Beilstein:
1299457
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

~50 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

COc1cc2OC3OC=CC3c2c4Oc5cccc(O)c5C(=O)c14

InChI

1S/C18H12O6/c1-21-11-7-12-13(8-5-6-22-18(8)24-12)17-15(11)16(20)14-9(19)3-2-4-10(14)23-17/h2-8,18-19H,1H3

InChI key

UTSVPXMQSFGQTM-UHFFFAOYSA-N

General description

Sterigmatocystin is classified under the mycotoxin family of compounds. It is usually produced by the fungi of several species of Aspergillus.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sterigmatocystin may be used as an analytical reference standard for the determination of the analyte in:
  • Grain samples by high-performance liquid chromatography-electrospray positive ionization-tandem mass spectrometry (HPLC-ESI+-MS/MS).
  • Beer samples by HPLC in conjunction with UV detection.

Analysis Note

purity : ≥96.0% (HPLC)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of sterigmatocystin in beer by high performance liquid chromatography with ultraviolet detection.
Ver?ilovskis A, et al.
World Mycotoxin Journal, 1(2), 161-166 (2008)
Analytical method for the determination of sterigmatocystin in grains using high-performance liquid chromatography-tandem mass spectrometry with electrospray positive ionization.
Ver?ilovskis A, et al.
Journal of Chromatography A, 1157(1-2), 467-471 (2007)
Odile Cabaret et al.
Rapid communications in mass spectrometry : RCM, 25(19), 2704-2710 (2011-09-14)
Mycotoxins are secondary metabolites of filamentous fungi which can cause a wide range of systemic effects. Human health effects of inhaled mycotoxins remain poorly documented, despite the large amounts present, associated with air-borne particles. Among these mycotoxins, sterigmatocystin is one
Daniela Jakšić et al.
Archives of toxicology, 86(10), 1583-1591 (2012-06-01)
Aspergillus versicolor and A. flavus are primary colonizers in damp dwellings, and they produce sterigmatocystin (ST) and aflatoxin B1 (AFB(1)), respectively. These hepatotoxic and carcinogenic mycotoxins and their precursors and derivates possess a furofuran ring, which has proven responsible for
C Huţanaşu et al.
Revista medico-chirurgicala a Societatii de Medici si Naturalisti din Iasi, 115(1), 33-37 (2011-06-22)
Sterigmatocystin (STC) is a wide spread mycotoxin produced by Aspergillus fungi, with hepatotoxic and carcinogenetic proprieties. To determine the STC concentration in blood and urine from patient with liver cirrhosis (LC) and hepatocellular carcinoma (HCC), with correlation with liver function

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service