Skip to Content
Merck
All Photos(1)

Documents

S100HR

Sigma-Aldrich

Sephacryl®

100-HR, MW range 1000-100,000 Da (globular proteins)

Synonym(s):

Poly([allyl dextran]-co-N,N′-methylenebisacrylamide)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56

Quality Level

description

stability: stable to 0.2M NaOH

form

suspension (20% aqueous ethanol)

feature

autoclavable

matrix

crosslinked allyl dextran/N,N′-methylenebisacrylamide copolymer

particle size

25-75 μm (wet)
25-75 μm (wet)

pore size

1000-100,000 Da MW range (globular proteins)

compatibility

mode of use size exclusion chromatography

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

  • Supplied swollen, suspended in 20% aqueous ethanol
  • Autoclavable
  • Stable to 0.2N sodium hydroxide (NaOH)
  • Store at refrigerated temperature (2-8 °C)

Application

Sephacryl® has been used:
  • to develop a novel high-throughput screening assay for sickle cell disease drug discovery
  • to purify and characterize ovine pancreatic elastase as well as a lysozyme from human urine
For size exclusion chromatography; the HR grade is a smaller particle size with narrower size range distribution, allowing for more efficient separations at faster flow rates. Recommended fractionation ranges are listed for globular proteins and dextrans..

Legal Information

Sephacryl is a registered trademark of Cytiva

related product

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

100.4 - 109.4 °F - closed cup

Flash Point(C)

38 - 43 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Novak et al.
Bioelectrochemistry (Amsterdam, Netherlands), 119, 136-141 (2017-10-11)
Cytochrome c (cyt c) is one of the most studied conjugated proteins due to its electron-transfer properties and ability to regulate the processes involved in homeostasis or apoptosis. Here we report an electrochemical strategy for investigating the electroactivity of cyt
Diederik Johannes Opperman et al.
Journal of bacteriology, 190(8), 3076-3082 (2008-02-12)
Bacteria can reduce toxic and carcinogenic Cr(VI) to insoluble and less toxic Cr(III). Thermus scotoductus SA-01, a South African gold mine isolate, has been shown to be able to reduce a variety of metals, including Cr(VI). Here we report the
Matthew Lord et al.
The Journal of cell biology, 167(2), 315-325 (2004-10-27)
We purified native Myo2p/Cdc4p/Rlc1p (Myo2), the myosin-II motor required for cytokinesis by Schizosaccharomyces pombe. The Myo2p heavy chain associates with two light chains, Cdc4p and Rlc1p. Although crude Myo2 supported gliding motility of actin filaments in vitro, purified Myo2 lacked
Marc A Benson et al.
Clinica chimica acta; international journal of clinical chemistry, 411(3-4), 190-197 (2009-11-11)
Pseudomonas aeruginosa is an opportunistic pathogen that causes disease in immunocompromised individuals, burn victims, and cystic fibrosis patients. Strains that secrete ExoU induce host cell lysis and damage epithelial tissue, which can lead to severe outcomes including sepsis and mortality.
S A Kashani et al.
Biomaterials, artificial cells, and immobilization biotechnology : official journal of the International Society for Artificial Cells and Immobilization Biotechnology, 19(3), 579-598 (1991-01-01)
We previously reported that the cell free supernatant of hepatocyte cultures contained a hepatic stimulatory factor, which increases survival time and stimulates liver regeneration in the galactosamine induced fulminant hepatic failure animal model. When hepatocytes were microencapsulated in an alginate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service