Skip to Content
Merck

Skip To

4B200

Sepharose 4B

45-165 μm bead diameter

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.21
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

algae (marine)

Quality Segment

form

suspension

technique(s)

affinity chromatography: suitable

bead diameter

45-165 μm

pore size

30,000-50,000 Da fractionation range (Dextrans), 60,000-20,000,000 Da fractionation range (Globular proteins)

storage temp.

2-8°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@H]([C@H]1CO)O)O[C@@H]4O[C@@H]5[C@H]([C@@H](OC5)[C@@H]4O)O[C@@H]6O[C@@H]([C@@H]([C@@H]([C@H]6O)O)O)CO)O)O[C@H]2[C@H]3OC[C@@H]2O[C@H]([C@H]3O)O

InChI

1S/C24H38O19/c25-1-5-9(27)11(29)12(30)22(38-5)41-17-8-4-36-20(17)15(33)24(40-8)43-18-10(28)6(2-26)39-23(14(18)32)42-16-7-3-35-19(16)13(31)21(34)37-7/h5-34H,1-4H2/t5-,6-,7+,8+,9+,10+,11+,12-,13+,14-,15+,16-,17-,18+,19+,20+,21-,22+,23+,24+/m1/s1

InChI key

MJQHZNBUODTQTK-WKGBVCLCSA-N

General description

Sepharose 4B is an agarose bead and a polymer of Gal β1,4 [3,6]-anhydro-L-galactose. Sepharose aids in fractionating larger molecules and is devoid of charged saccharides. Presence of 3,6-anhydro-L-galactose resists biological degradation. Hydrogen bonds stabilize the sepharose gel structure. Sepharose is widely used in affinity chromatography by cross linking various ligands on the matrix.

Application

Sepharose 4B has been used:
  • to conjugate scorpion venom to be used in affinity chromatography
  • for clearing of protein extracts prior to immunoprecipitation
  • for gel purification and desalting of phospholipid vesicles
  • for adsorption of immunoglobulin G (IgG)[1]

Legal Information

Sepharose is a trademark of Cytiva

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GE17-0120-01GE17-0110-016B100
technique(s)

affinity chromatography: suitable

technique(s)

-

technique(s)

-

technique(s)

affinity chromatography: suitable

biological source

algae (marine)

biological source

-

biological source

-

biological source

algae (marine)

Quality Level

200

Quality Level

-

Quality Level

-

Quality Level

-

form

suspension

form

-

form

-

form

suspension

storage temp.

2-8°C

storage temp.

-

storage temp.

-

storage temp.

2-8°C

pore size

30,000-50,000 Da fractionation range (Dextrans), 60,000-20,000,000 Da fractionation range (Globular proteins)

pore size

-

pore size

-

pore size

10,000-1,000,000 Da fractionation range (dextrans), 10,000-4,000,000 Da fractionation range (globular proteins)


Still not finding the right product?

Explore all of our products under Sepharose 4B


pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

100.4 - 109.4 °F - closed cup

flash_point_c

38 - 43 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters