Skip to Content
Merck

Skip To

DCL6B100

DEAE–Sepharose

CL-6B

Synonym(s):

Diethylaminoethyl–Sepharose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
47101511
NACRES:
NA.56
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

suspension

technique(s)

affinity chromatography: suitable

matrix

6% cross-linked agarose

bead size

45-165 μm

pore size

~4,000,000 Da exclusion limit

pH

3—12

capacity

130-170 μeq/mL binding capacity (gel volume)(gel volume)

General description

DCL6B100-500ML′s updated product number is GE17-0710-01

Application

DEAE-Sepharose® is used in affinity chromatography, protein chromatography and ion exchange chromatography. DEAE-Sepharose has been used to study pathogenesis of human disease and to develop a new assay for detecting the toxins of pathogenic strains of Clostridium difficile.

Legal Information

DEAE-Sepharose is a registered trademark of Cytiva
Sepharose is a trademark of Cytiva

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
DFF100A50120CCF100
pore size

~4,000,000 Da exclusion limit

pore size

~4,000,000 Da exclusion limit

pore size

~200,000 Da exclusion limit

pore size

4,000,000 exclusion limit (av. mol. wt.)

matrix

6% cross-linked agarose

matrix

6% cross-linked agarose

matrix

cross-linked dextran

matrix

6% cross-linked agarose

capacity

130-170 μeq/mL binding capacity (gel volume)(gel volume)

capacity

110-160 μeq/mL binding capacity(gel volume)

capacity

3-4 meq/g binding capacity

capacity

90-130 μeq/mL(gel basis)

technique(s)

affinity chromatography: suitable

technique(s)

affinity chromatography: suitable

technique(s)

protein purification: suitable

technique(s)

affinity chromatography: suitable

form

suspension

form

suspension

form

powder

form

suspension

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200


Still not finding the right product?

Explore all of our products under DEAE–Sepharose


pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

100.4 - 109.4 °F

flash_point_c

38 - 43 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters