Skip to Content
Merck
All Photos(2)

Key Documents

P8293

Sigma-Aldrich

Protoporphyrin IX

≥95%

Synonym(s):

3,7,12,17-Tetramethyl-8,13-divinyl-2,18-porphinedipropionic acid, Kammerer’s porphyrin, Ooporphyrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H34N4O4
CAS Number:
Molecular Weight:
562.66
Beilstein:
380795
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.32

biological source

Porcine

Quality Level

Assay

≥95%

form

powder

solubility

DMF: methanol (1:1): soluble-10 mg/mL, dark red-brown

fluorescence

λex 402 nm; λem 625 nm (bound to HRP)
λex 409 nm; λem 633 nm

storage temp.

2-8°C

SMILES string

Cc1c(CCC(O)=O)c2cc3[nH]c(cc4nc(cc5[nH]c(cc1n2)c(C)c5C=C)c(C)c4C=C)c(C)c3CCC(O)=O

InChI

1S/C34H34N4O4/c1-7-21-17(3)25-13-26-19(5)23(9-11-33(39)40)31(37-26)16-32-24(10-12-34(41)42)20(6)28(38-32)15-30-22(8-2)18(4)27(36-30)14-29(21)35-25/h7-8,13-16,35,38H,1-2,9-12H2,3-6H3,(H,39,40)(H,41,42)/b25-13-,26-13-,27-14-,28-15-,29-14-,30-15-,31-16-,32-16-

InChI key

KSFOVUSSGSKXFI-UJJXFSCMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Protoporphyrin IX belongs to the porphyrin family, which are a class of tetrapyrroles. It is the iron-free form of hemin and amphiphilic in nature. It is the precursor of heme in its biosynthetic pathway.
Protoporphyrin IX imparts red color to heme and is the major porphyrin in blood. It is synthesized from protoporphyrinogen IX in the presence of protoporphyrinogen IX oxidase.

Application

Activates soluble guanylyl cyclase.
Protoporphyrin IX has been used:
  • as a standard in protoporphyrin assays
  • in fluorescence spectra analysis
  • to treat cells in cell culture to study heme-mediated ferroportin 1 transcription

Biochem/physiol Actions

Protoporphyrin IX penetrates human erythrocytes and aids the release of oxygen from them. It forms complexes with myoglobin and hemoglobin. It is a photosensitizer used in the photodynamic therapy of cancer.
Protoporphyrin IX levels are elevated in tumor cells due to metabolism anomalies compared to normal cells.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feng Liu et al.
Sensors (Basel, Switzerland), 18(6) (2018-06-03)
A label-free sensing platform is developed based on switching the structure of aptamer for highly sensitive and selective fluorescence detection of ochratoxin A (OTA). OTA induces the structure of aptamer, transforms into G-quadruplex and produces strong fluorescence in the presence
Heme controls ferroportin1 (FPN1) transcription involving Bach1, Nrf2 and a MARE/ARE sequence motif at position -7007 of the FPN1 promoter.
Marro S
Haematologica, 95(8), 1261-1268 (2010)
Ki Mo Kim et al.
Environmental toxicology and pharmacology, 48, 85-93 (2016-10-23)
Coniferaldehyde (CA) exerts anti-inflammatory properties by inducing heme oxygenase-1 (HO-1). To define the regulation mechanism by which CA induces a cytoprotective function and HO-1 expression, the up-stream regulations involved in the activation of nuclear transcription factor-erythroid 2-related factor (Nrf)-2/HO-1 pathway
J Flemmig et al.
Free radical research, 50(12), 1386-1395 (2016-10-26)
Excessive release of hemoglobin from red blood cells markedly disturbs the health status of patients due to cytotoxic effects of free hemoglobin and heme. The latter component is able to initiate novel hemolytic events in unperturbed red blood cells. We
Intrinsic fluorescence of protoporphyrin IX from blood samples can yield information on the growth of prostate tumours
de Oliveira Silva FR, et al.
Journal of Fluorescence, 20(6), 1159-1165 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service