Skip to Content
Merck
All Photos(2)

Key Documents

P2076

Sigma-Aldrich

Polymyxin B nonapeptide hydrochloride

lyopholized powder

Synonym(s):

PMBN

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H74N14O11 · xHCl
Molecular Weight:
963.13 (free base basis)
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

Polymyxin B nonapeptide hydrochloride, cationic cyclic peptide

Quality Level

form

lyophilized powder

color

white to light yellow

application(s)

cell analysis

storage temp.

2-8°C

SMILES string

CC(C)CC1NC(=O)C(Cc2ccccc2)NC(=O)C(CCN)NC(=O)C(CCNC(=O)C(NC(=O)C(CCN)NC(=O)C(CCN)NC1=O)C(C)O)NC(=O)C(CCN)NC(=O)C(N)C(C)O

InChI

1S/C43H74N14O11/c1-22(2)20-31-40(65)52-26(10-15-44)35(60)51-29(13-18-47)39(64)57-34(24(4)59)43(68)49-19-14-30(53-36(61)28(12-17-46)54-42(67)33(48)23(3)58)38(63)50-27(11-16-45)37(62)56-32(41(66)55-31)21-25-8-6-5-7-9-25/h5-9,22-24,26-34,58-59H,10-21,44-48H2,1-4H3,(H,49,68)(H,50,63)(H,51,60)(H,52,65)(H,53,61)(H,54,67)(H,55,66)(H,56,62)(H,57,64)

InChI key

PYHYGIPVYYRJHU-UHFFFAOYSA-N

Application

Polymyxin B nonapeptide hydrochloride has been used as an outer membrane permeabilizer, to characterize PAßN activity on membrane. It has been used as a control in antibacterial assays.

Biochem/physiol Actions

PMBN has endotoxin-neutralizing activity and thus can be utilized in adjunctive therapy against Gram-negative sepsis. It has been found that PMBN is less toxic than polymyxin B. Unlike polymyxin B, PMBN does not exhibit neurotoxicity and nephrotoxicity. While it retains the anti endotoxin property of the parent compound, it is much less potent.
Polymyxin B nonapeptide (PMBN), a cationic cyclic peptide derived from the antibacterial peptide polymyxin B, specifically increases the permeability of the outer membrane of Gram-negative bacteria toward hydrophobic antibiotics. PMBN has been used to evaluate multidrug efflux inhibitors in E. coli.
Polymyxin B nonapeptide hydrochloride is a derivative of PMB that induces outer membrane permeability in gram-negative bacteria.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Vaara
Microbiological reviews, 56(3), 395-411 (1992-09-01)
The outer membrane of gram-negative bacteria provides the cell with an effective permeability barrier against external noxious agents, including antibiotics, but is itself a target for antibacterial agents such as polycations and chelators. Both groups of agents weaken the molecular
Hongfei Pi et al.
Frontiers in microbiology, 11, 1556-1556 (2020-08-28)
Multidrug-resistant (MDR) pathogens, particularly the ESKAPE group (Enterococcus faecalis/faecium, Staphylococcus aureus, Klebsiella pneumoniae, Acinetobacter baumannii, Pseudomonas aeruginosa, Escherichia coli, and Enterobacter spp.), have become a public health threat worldwide. Development of new antimicrobial classes and the use of drugs in
Purification, toxicity, and antiendotoxin activity of polymyxin B nonapeptide.
Danner RL
Antimicrobial Agents and Chemotherapy, 33(9), 1428-1434 (1989)
Yoshimi Matsumoto et al.
PloS one, 6(4), e18547-e18547 (2011-05-03)
Fluorescein-di-β-D-galactopyranoside (FDG), a fluorogenic compound, is hydrolyzed by β-galactosidase in the cytoplasm of Escherichia coli to produce a fluorescent dye, fluorescein. We found that both FDG and fluorescein were substrates of efflux pumps, and have developed a new method to
Evaluation of multidrug efflux pump inhibitors by a new method using microfluidic channels.
Matsumoto Y
PLoS ONE, 6(4), 1-12 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service