Skip to Content
Merck
All Photos(2)

Key Documents

P1004

Sigma-Aldrich

Polymyxin B sulfate salt

Synonym(s):

Polymixin, Polymixin B, Polymixin B sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C55H96N16O13 · 2H2SO4
CAS Number:
Molecular Weight:
1385.61
EC Number:
MDL number:
UNSPSC Code:
51283106
NACRES:
NA.76

biological source

Pseudomonas aeruginosa

Quality Level

form

powder

storage condition

(Tightly closed. Dry. )

solubility

water: 50 mg/mL

antibiotic activity spectrum

Gram-negative bacteria
fungi

Mode of action

cell membrane | interferes

storage temp.

2-8°C

SMILES string

O=C(C(NC(C(CCN)NC(CCCCC(C)CC)=O)=O)C(C)O)NC(CCN)C(NC(CCNC(C(NC(C(NC(C(CCN)N1)=O)CCN)=O)C(O)C)=O)C(NC(CCN)C(NC(CC2=CC=CC=C2)C(NC(CC(C)C)C1=O)=O)=O)=O)=O.O=S(O)(O)=O

InChI

1S/C48H82N16O13.H2O4S/c1-27(2)24-37-47(76)59-32(11-19-52)41(70)56-31(10-18-51)43(72)61-35(14-22-65)39(68)54-21-13-34(45(74)57-33(12-20-53)44(73)64-38(48(77)63-37)25-28-6-4-3-5-7-28)60-42(71)30(9-17-50)58-46(75)36(15-23-66)62-40(69)29(8-16-49)55-26-67;1-5(2,3)4/h3-7,26-27,29-38,65-66H,8-25,49-53H2,1-2H3,(H,54,68)(H,55,67)(H,56,70)(H,57,74)(H,58,75)(H,59,76)(H,60,71)(H,61,72)(H,62,69)(H,63,77)(H,64,73);(H2,1,2,3,4)

InChI key

HNDFYNOVSOOGDU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polymyxin B sulfate is a strongly cationic cyclic polypeptide antibiotic that is derived from fermentation of Bacilus polymyxa. It is a mixture of B1 and B2 sulfate.

Application

Polymyxin B sulfate is a strongly cationic cyclic polypeptide antibiotic that is derived from fermentation of Bacilus polymyxa. It is a mixture of B1 and B2 sulfate. The product has been used clinically to treat infections of the urinary tract, meninges and blood stream caused by susceptible strains of Pseudomonas aeruginosa. It also has uses studying multidrug-resistant pathogens††, as an immobilized agent for removal of endotoxins, and to induce pore formation in the membranes of cortex cells from excised sorghum roots.
Polymyxin B sulfate salt has been used:
  • in human aortic endothelial cells (HAEC) cell culture to exclude potential endotoxin contamination in recombinant adiponectin
  • in the analysis of the isolates
  • in endotoxin-neutralizing activity of α-amylase (AmyI-1)-18 (LAL assay)

Biochem/physiol Actions

Mode of Action: Polymyxin B Sulfate binds to the lipid A portion of bacterial lipopolysaccharides†, disrupting the cytoplasmic membrane by inducing pores large enough to permit nucleotide leakage in bacterial walls. This disrupts the permeability of the cytoplasmic membrane.

Antimicrobial spectrum: Has bactericidal action on most gram-negative bacilli††, including E. Coli and on most fungi and gram-positive bacteria.
Polymyxin B is one of the most important antibiotics used to treat gram-negative bacterial infections, including infections caused by carbapenem-resistant non-fermenters and carbapenem-resistant Enterobacteriaceae.

Features and Benefits

High quality antibiotic suitable for mulitple research applications

Preparation Note

Polymyxin B sulfate is soluble in water at 50 mg/mL, producing a very slightly hazy, colorless to yellow solution. It has only minimal solubility in any organic solvent. For example, it has a solubility of 0.115 mg/mL in ethanol.

Stock solutions should be sterile filtered and stored at 2-8°C. They are stable at 37°C for 5 days.

Storage and Stability

As supplied, the product should be stored at 2-8°C in the dark. No change was observed in retained samples after three years′ of storage in these conditions.

Other Notes

For additional information on our range of Biochemicals, please complete this form.
Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Endotoxin-neutralizing activity and mechanism of action of a cationic alpha-helical antimicrobial octadecapeptide derived from alpha-amylase of rice
Taniguchi M, et al.
Peptides, 101-108 (2016)
Dosing and pharmacokinetics of polymyxin B in patients with renal insufficiency
Thamlikitkul V, et al.
Antimicrobial Agents and Chemotherapy, e01337-e01316 (2017)
Biochemical and toxic diversity of Bacillus cereus in a pasta and meat dish associated with a food-poisoning case
Pirhonen T I, et al.
Food Microbiology, 22(1), 87-91 (2005)
Globular adiponectin counteracts VCAM-1-mediated monocyte adhesion via AdipoR1/NF-kappaB/COX-2 signaling in human aortic endothelial cells
Addabbo F, et al.
American Journal of Physiology. Endocrinology and Metabolism, 301(6), E1143-E1143 (2011)
Xucheng Huang et al.
International journal of medical microbiology : IJMM, 308(7), 819-828 (2018-07-24)
Ureaplasma spp. are known to be associated with human genitourinary tract diseases and perinatal diseases and Ureaplasma spp. Lipid-associated membrane proteins (LAMPs) play important roles in their related diseases. However, the exact mechanism underlying pathogenesis of Ureaplasma spp. LAMPs is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service