Skip to Content
Merck
All Photos(3)

Key Documents

N7778

Sigma-Aldrich

7-Nitroindazole

≥98%

Synonym(s):

7-NI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5N3O2
CAS Number:
Molecular Weight:
163.13
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98%

form

powder

solubility

DMF: 50 mg/mL, clear, yellow to orange

storage temp.

−20°C

SMILES string

[O-][N+](=O)c1cccc2cn[nH]c12

InChI

1S/C7H5N3O2/c11-10(12)6-3-1-2-5-4-8-9-7(5)6/h1-4H,(H,8,9)

InChI key

PQCAUHUKTBHUSA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Selective inhibitor of nitric oxide synthase from mouse brain.

Features and Benefits

This compound is featured on the Nitric Oxide Synthases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Philippe Bonnin et al.
International journal of molecular sciences, 21(18) (2020-09-12)
The poly(ADP-ribose) polymerase (PARP) inhibitor PJ34 has been reported to improve endothelial dysfunction in the peripheral system. We addressed the role of PJ34 on the vascular tone and vasoreactivity during development in the mouse brain. Blood flows were measured in
Geoffrey E Ciarlone et al.
American journal of physiology. Cell physiology, 311(6), C1014-C1026 (2016-10-14)
Central CO2-chemosensitive neurons in the caudal solitary complex (cSC) are stimulated not only by hypercapnic acidosis, but by hyperoxia as well. While a cellular mechanism for the CO2 response has yet to be isolated, previous data show that a redox-sensitive
Morten Hedetoft et al.
Undersea & hyperbaric medicine : journal of the Undersea and Hyperbaric Medical Society, Inc, 45(3), 335-350 (2018-07-22)
Experiments have shown that hyperbaric oxygen (HBO2) therapy reduces cyanide-induced cerebral distress. The exact mechanism behind HBO2's neuroprotective effect is unknown, but has been proposed to be mediated by an increased neuronal nitric oxide (NO) bioavailability, which may compete with
Anita J Turner et al.
American journal of physiology. Renal physiology, 313(4), F864-F873 (2017-07-07)
Via developmental programming, prenatal perturbations, such as exposure to glucocorticoids and maternal malnutrition alter kidney development and contribute to the development of hypertension. To examine the possibility that alterations in tubuloglomerular feedback (TGF) contribute to the development of hypertension in
J Garthwaite
Trends in neurosciences, 14(2), 60-67 (1991-02-01)
Nitric oxide (NO) is a recently discovered and highly unorthodox messenger molecule. Current evidence indicates that, in the CNS, NO is produced enzymatically in postsynaptic structures in response to activation of excitatory amino acid receptors. It then diffuses out to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service