Skip to Content
Merck
All Photos(1)

Key Documents

N5501

Sigma-Aldrich

Nω-Nitro-L-arginine

≥98% (TLC)

Synonym(s):

L-NNA, N5-(Nitroamidino)-L-2,5-diaminopentanoic acid, NG-NO2-L-Arg

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13N5O4
CAS Number:
Molecular Weight:
219.20
Beilstein:
1728914
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

Assay

≥98% (TLC)

form

powder

mp

257 °C

solubility

1 M HCl: 50 mg/mL, clear, colorless to faintly yellow

SMILES string

N[C@@H](CCCNC(=N)N[N+]([O-])=O)C(O)=O

InChI

1S/C6H13N5O4/c7-4(5(12)13)2-1-3-9-6(8)10-11(14)15/h4H,1-3,7H2,(H,12,13)(H3,8,9,10)/t4-/m0/s1

InChI key

MRAUNPAHJZDYCK-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Nω-Nitro-L-arginine has been used:
  • as nitric oxide synthase (NOS) inhibitor, to measure the release of nitric oxide (NO) derivatives in cytochrome c reduction assay
  • to pretreat fiber bundles for contractile studies
  • as NOS inhibitor to characterize the generation of NO in Taxus yunnanensis cells
  • as an irreversible inhibitor of the neuronal NO synthase (nNOS) to preincubate swimbladder preparations

Biochem/physiol Actions

Nω-Nitro-L-arginine/L-NNA helps to reduce the levels of cortical hyperemia and brain cGMP occurring due to hypercapnia.
Irreversible inhibitor of constitutive nitric oxide synthase (nNOS) and a reversible inhibitor of inducible nitric oxide synthase (iNOS).

Features and Benefits

This compound is featured on the Nitric Oxide Synthases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nitric oxide modulates excitation-contraction coupling in the diaphragm
Reid M B, et al.
Comparative Biochemistry and Physiology. Part A, Molecular & Integrative Physiology, 119(1), 211-218 (1998)
Helena Janols et al.
Journal of leukocyte biology, 96(5), 685-693 (2014-06-15)
The causative microorganisms dictate the type of MDSC generated in sepsis patients, and a large proportion of PMN-MDSCs in gram-positive sepsis includes immunosuppressive myeloid blasts. MDSCs constitute a heterogeneous population of immature myeloid cells that potently suppress immune responses. They
K D Keef et al.
The Journal of physiology, 591(6), 1489-1506 (2013-01-23)
There is evidence that vasoactive intestinal polypeptide (VIP) participates in inhibitory neuromuscular transmission (NMT) in the internal anal sphincter (IAS). However, specific details concerning VIP-ergic NMT are limited, largely because of difficulties in selectively blocking other inhibitory neural pathways. The
Involvement of nitric oxide in oxidative burst, phenylalanine ammonia-lyase activation and Taxol production induced by low-energy ultrasound in Taxus yunnanensis cell suspension cultures.
Wang JW
Nitric Oxide, 15(4), 351-358 (2006)
Ming-Guo Feng et al.
American journal of physiology. Renal physiology, 303(5), F775-F782 (2012-06-08)
Nebivolol is a β(1)-adrenergic blocker that also elicits renal vasodilation and increases the glomerular filtration rate (GFR). However, its direct actions on the renal microvasculature and vasodilator mechanism have not been established. We used the in vitro blood-perfused juxtamedullary nephron

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service