Skip to Content
Merck
All Photos(1)

Key Documents

M7949

Sigma-Aldrich

Mitomycin C from Streptomyces caespitosus

≥98% (HPLC), γ-irradiated, suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18N4O5
CAS Number:
Molecular Weight:
334.33
Beilstein:
7231816
EC Number:
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85

biological source

Streptomyces caespitosus

Quality Level

sterility

γ-irradiated

Assay

≥98% (HPLC)

form

powder or crystals

technique(s)

cell culture | mammalian: suitable

color

dark blue

mp

>360 °C

solubility

DMSO: 1.0 mg/mL

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
neoplastics

Mode of action

DNA synthesis | interferes

storage temp.

2-8°C

SMILES string

[H][C@]12CN3C4=C([C@@H](COC(N)=O)[C@@]3(OC)[C@@]1([H])N2)C(=O)C(N)=C(C)C4=O

InChI

1S/C15H18N4O5/c1-5-9(16)12(21)8-6(4-24-14(17)22)15(23-2)13-7(18-13)3-19(15)10(8)11(5)20/h6-7,13,18H,3-4,16H2,1-2H3,(H2,17,22)/t6-,7+,13+,15-/m1/s1

InChI key

NWIBSHFKIJFRCO-WUDYKRTCSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Mitomycin C is produced by a strain of actinomyces, Streptomyces caespitosus. It contains three anticancer moieties, quinine, urethane, and aziridine groups. It is used to generate mitotically inactive feeder cells in cell culture systems, such as the mitotically inactive fibroblasts used in embryonic stem cell systems.

Biochem/physiol Actions

Mode of Action: This product is an alkylating agent that specifically targets the guanine nucleoside sequence 5′-CpG-3′. It inhibits DNA synthesis by covalently reacting with DNA, forming crosslinks between complementary strands of DNA. This interaction prevents separation of complementary DNA strands, inhibiting DNA replication.

Antimicrobial Spectrum: Mitomycin C has strong antitumor activity, especially against Ehrlich ascites tumor cells, and strong bactericidal action against gram-positive and gram-negative bacteria.

Caution

Stock solutions should be filter sterilized and stored at 2-8°C in the dark. Solutions at pH 6-9 can be stored at 0-5°C for up to a week, but if a precipitate forms, a fresh solution should be prepared - the precipitated solution has been proven toxic to cells.

Preparation Note

Mitomycin C is soluble in DMSO at 1.0 mg/mL

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Carc. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yardena Tenenbaum-Rakover et al.
Journal of medical genetics, 52(6), 391-399 (2015-04-16)
Primary gonadal failure is characterised by primary amenorrhoea or early menopause in females, and oligospermia or azoospermia in males. Variants of the minichromosome maintenance complex component 8 gene (MCM8) have recently been shown to be significantly associated with women's menopausal
Xianbin Zhang et al.
Journal of Cancer, 11(2), 479-487 (2020-01-04)
The poor survival rate of pancreatic cancer is still a major challenge for the clinicians and their patients. In this study, we evaluated the efficacy of metformin, an inhibitor of oxidative phosphorylation, in combination with LW6, which impairs malate dehydrogenase
Annika Gillis et al.
Applied and environmental microbiology, 80(14), 4138-4152 (2014-05-06)
GIL01, Bam35, GIL16, AP50, and Wip1 are tectiviruses preying on the Bacillus cereus group. Despite the significant contributions of phages in different biological processes, little is known about the dealings taking place between tectiviruses and their Gram-positive bacterial hosts. Therefore
Sayuri Yamazaki et al.
Journal of immunology (Baltimore, Md. : 1950), 193(11), 5488-5497 (2014-10-29)
Accumulating evidence shows that immunological tolerance induced by Ag administration together with UVB irradiation is dependent on Foxp3(+) CD4(+) regulatory T (Treg) cells. However, the mechanisms by which UVB controls Treg cells in the skin are currently unclear. In this
Marcony R Santhiago et al.
Cornea, 31(3), 311-321 (2011-12-14)
To provide an overview of the safety and efficacy of mitomycin C (MMC) as adjuvant therapy after refractive surgery procedures. Literature review. Over the past 10 years, MMC has been used by refractive surgeons to prophylactically decrease haze after surface

Articles

Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic

Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic

Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic

Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service