Skip to Content
Merck

C2618

Cytochalasin D

Ready Made Solution, from Zygosporium mansonii, 5 mg/mL in DMSO

Synonym(s):

Lygosporin A, Zygosporin A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C30H37NO6
CAS Number:
Molecular Weight:
507.62
UNSPSC Code:
51102829
NACRES:
NA.85
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1632828
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Zygosporium mansonii

Quality Segment

form

ready-to-use solution

concentration

5 mg/mL in DMSO

antibiotic activity spectrum

fungi

application(s)

advanced drug delivery

mode of action

enzyme | interferes

shipped in

wet ice

storage temp.

−20°C

SMILES string

[H][C@@]12[C@H](C)C(=C)[C@@H](O)[C@@H]3\C=C\C[C@H](C)C(=O)[C@](C)(O)\C=C\[C@@H](OC(C)=O)[C@@]13C(=O)N[C@H]2Cc4ccccc4

InChI

1S/C30H37NO6/c1-17-10-9-13-22-26(33)19(3)18(2)25-23(16-21-11-7-6-8-12-21)31-28(35)30(22,25)24(37-20(4)32)14-15-29(5,36)27(17)34/h6-9,11-15,17-18,22-26,33,36H,3,10,16H2,1-2,4-5H3,(H,31,35)/b13-9+,15-14+/t17-,18+,22-,23-,24+,25-,26+,29+,30+/m0/s1

InChI key

SDZRWUKZFQQKKV-JHADDHBZSA-N

Application

Cytochalasin D are isomeric metabolites of Metarrhizium anisopliae. Cytochalasin D possesses antibiotic and antitumor activity. It also impairs maintenance of long term potentiation (LTP) of actin filaments. It is implicated in promoting conditions favorable for depolymerizing actin.

Biochem/physiol Actions

Cell permeable fungal toxin; potent inhibitor of actin polymerization. Disrupts actin microfilaments and activates the p53-dependent pathways causing arrest of the cell cycle at the G1-S transition. Inhibits smooth muscle contraction. Inhibits insulin-stimulated, but not basal, transport of glucose.
Potent inhibitor of actin polymerization; disrupts actin microfilaments; activates the p53-dependent pathways; inhibits smooth muscle contraction; inhibits insulin-stimulated glucose transport.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.


Still not finding the right product?

Explore all of our products under Cytochalasin D


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

188.6 °F - closed cup - Solvent

flash_point_c

87 °C - closed cup - Solvent

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library