Skip to Content
Merck
All Photos(2)

Documents

M0440

Sigma-Aldrich

Mitomycin C from Streptomyces caespitosus

meets USP testing specifications

Synonym(s):

Mitomycin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18N4O5
CAS Number:
Molecular Weight:
334.33
Beilstein:
7231816
EC Number:
MDL number:
UNSPSC Code:
12352203
PubChem Substance ID:
NACRES:
NA.76

biological source

Streptomyces caespitosus

color

blue-violet

solubility

H2O: 40 mg/mL, clear to very slightly hazy, colorless
H2O: soluble

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

DNA synthesis | interferes

storage temp.

2-8°C

SMILES string

[H][C@]12CN3C4=C([C@@H](COC(N)=O)[C@@]3(OC)[C@@]1([H])N2)C(=O)C(N)=C(C)C4=O

InChI

1S/C15H18N4O5/c1-5-9(16)12(21)8-6(4-24-14(17)22)15(23-2)13-7(18-13)3-19(15)10(8)11(5)20/h6-7,13,18H,3-4,16H2,1-2H3,(H2,17,22)/t6-,7+,13+,15-/m1/s1

InChI key

NWIBSHFKIJFRCO-WUDYKRTCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: aziridine

Application

Mitomycin C is produced by a strain of actinomyces, Streptomyces caespitosus. It contains three anticancer moieties, quinine, urethane, and aziridine groups. It is used to generate mitotically inactive feeder cells in cell culture systems, such as the mitotically inactive fibroblasts used in embryonic stem cell systems.

Biochem/physiol Actions

Mode of Action: This product is an alkylating agent that specifically targets the guanine nucleoside sequence 5′-CpG-3′. It inhibits DNA synthesis by covalently reacting with DNA, forming crosslinks between complementary strands of DNA. This interaction prevents separation of complementary DNA strands, inhibiting DNA replication.

Antimicrobial Spectrum: Mitomycin C has strong antitumor activity, especially against Ehrlich ascites tumor cells, and strong bactericidal action against gram-positive and gram-negative bacteria.

Caution

Stock solutions should be filter sterilized and stored at 2-8 °C in the dark. Solutions at pH 6-9 can be stored at 0-5 °C for up to a week, but if a precipitate forms, a fresh solution should be prepared - the precipitated solution has been proven toxic to cells.

Preparation Note

Mitomycin C is soluble in water at .5 mg/mL, with a pH of 6.0-7.5. It undergoes rapid degradation in acidic solutions with pH<6, and is mostly likely to retain activity in solutions with a pH between 6-9.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral - Carc. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leonard L Gunderson et al.
International journal of radiation oncology, biology, physics, 87(4), 638-645 (2013-09-17)
The long-term update of US GI Intergroup RTOG 98-11 anal cancer trial found that concurrent chemoradiation (CCRT) with fluorouracil (5-FU) plus mitomycin had a significant impact on disease-free survival (DFS) and overall survival (OS) compared with induction plus concurrent 5-FU plus
Scott C Jeffrey et al.
Journal of medicinal chemistry, 48(5), 1344-1358 (2005-03-04)
Antibody-drug conjugates (ADCs) were prepared consisting of DNA minor groove binder drugs (MGBs) attached to monoclonal antibodies (mAbs) through peptide linkers designed to release drugs inside the lysosomes of target cells. The site of linker attachment on the MGB was
Xin Wang et al.
Nature medicine, 19(4), 473-480 (2013-03-26)
Sorting nexin 27 (SNX27), a brain-enriched PDZ domain protein, regulates endocytic sorting and trafficking. Here we show that Snx27(-/-) mice have severe neuronal deficits in the hippocampus and cortex. Although Snx27(+/-) mice have grossly normal neuroanatomy, we found defects in
Annika Gillis et al.
Applied and environmental microbiology, 80(14), 4138-4152 (2014-05-06)
GIL01, Bam35, GIL16, AP50, and Wip1 are tectiviruses preying on the Bacillus cereus group. Despite the significant contributions of phages in different biological processes, little is known about the dealings taking place between tectiviruses and their Gram-positive bacterial hosts. Therefore
Lars Petter Jordheim et al.
Molecular pharmacology, 84(1), 12-24 (2013-04-13)
The benefit of cancer chemotherapy based on alkylating agents is limited because of the action of DNA repair enzymes, which mitigate the damage induced by these agents. The interaction between the proteins ERCC1 and XPF involves two major components of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service