Skip to Content
Merck
All Photos(1)

Key Documents

I1408

Sigma-Aldrich

IMAC-Select Affinity Gel

(1:1 suspension in a 30% ethanol solution)

Synonym(s):

Affinity Gel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

form

(1:1 suspension in a 30% ethanol solution)

shelf life

1 yr

technique(s)

affinity chromatography: suitable

impurities

≤0.08 Metal Contamination (μmol/ml packed gel)

matrix

6% beaded agarose

capacity

10-30 μmol/mL binding capacity (Cu+2)

shipped in

wet ice

storage temp.

2-8°C

Application

An immobilized metal affinity chromatography (IMAC) product that can be widely used for purifying proteins, peptides, and nucleotides of interest, most commonly proteins. IMAC-Select utilizes a non-charged linkage chemistry to connect the proprietary quadridentate chelate to the agarose bead matrix. IMAC-Select Affinity Gel is durable and can capture proteins of interest with an affinity to various metal ions such as nickel, cobalt, iron or gallium. A metal free resin allows researchers to add metal of choice, or to use the resin to remove metal contamination from a protein. IMAC-Select Affinity Gel has been used in the affinity purification of kelch-like ECH-associated protein 1 (KEAP1), Wilms tumor gene on the X chromosome (WTX) and glycoprotein G1 tail protein of the andes virus.

Physical form

1:1 suspension in a 30% ethanol solution

Preparation Note

The ethanol used to store the affiinity gel must be removed prior to use to avoid precipitation of buffer salts.The affinity gel is first washed with 1 to 2 volumes of deionized water to remove ethanol. The gel is charged with 2 to 3 volumes of a solution of the desired metal ion at a concentration of 10 mg/mL, and then equilibrated with 3 to 5 volumes of equilibration buffer.

Other Notes

Do not allow the affinity gel to remain in any buffer for extended periods of time unless it contains an anti-microbial agent such as 30% ethanol.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The hantavirus glycoprotein G1 tail contains dual CCHC-type classical zinc fingers
Estrada DF, et al.
The Journal of Biological Chemistry, 284(13), 8654-8660 (2009)
Wilms tumor gene on the X chromosome (WTX) inhibits the degradation of NRF2 through competitive binding to KEAP1
Camp ND, et al.
The Journal of Biological Chemistry, jbc-M111 (2012)
Hao Li et al.
Proteomics, 9(6), 1646-1661 (2009-03-03)
Ethylene regulates a variety of stress responses and developmental adaptation in plants. In the present study, the phosphoproteomics is adopted to investigate the differential protein phosphorylation by ethylene in Arabidopsis ethylene-insensitive 2 (ein2) mutant. A total of 224 phosphopeptides were
Kris Gevaert et al.
Methods in molecular biology (Clifton, N.J.), 527, 219-227 (2009-02-26)
We present a gel-free proteomics procedure for the specific isolation of phosphorylated peptides from whole proteome digests. Central is the use of diagonal, reverse-phase chromatography which consists of two consecutive reverse-phase peptide separations with a modification step in between. The
Wilms tumor gene on the X chromosome (WTX) inhibits the degradation of NRF2 through competitive binding to KEAP1
Camp ND, et al.
The Journal of biological chemistry, jbc-M111 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service