Skip to Content
Merck
All Photos(1)

Documents

P9740

Sigma-Aldrich

PHOS-Select Iron Affinity Gel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

form

suspension

shelf life

2 yr at −20‑0 °C

pH

~5

capacity

≥2 μmol/mL, gel (phosphopeptide)(following a 30 minute incubation at 25°C)

storage temp.

−20°C

General description

The PHOS-Select iron affinity gel is a novel iron [Fe(III)] chelate matrix. This matrix provides high capacity affinity binding of molecules containing phosphate groups.

Application

Phos-Select products facilitate a number of applications including:
  • affinity enrichment of phosphopeptides obtained from protein tryptic digests
  • direct transfer of phosphocompounds for analysis (HPLC or mass spectrometry)
  • enrichment of phosphopeptides by immobilized metal affinity chromatography (IMAC)

Other Notes

Phos-Select Iron Affinity Gel is a novel iron [Fe(III)] chelate matrix based on Sigma′s NTA analog chelate ligand. The matrix provides high capacity affinity binding of molecules containing phosphate groups.

Physical form

aqueous glycerol suspension

Legal Information

PHOS-Select is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mathilde Biola-Clier et al.
Nanomaterials (Basel, Switzerland), 10(2) (2020-01-25)
TiO2 nanoparticles (NPs) are one of the most produced NPs worldwide and are used in many consumer products. Their impact on human health, especially through inhalation, has been studied for more than two decades. TiO2 is known for its strong
Jie Gao et al.
Scientific reports, 7, 40852-40852 (2017-01-21)
Low temperature is one of the key environmental stresses, which greatly affects global banana production. However, little is known about the global phosphoproteomes in Musa spp. and their regulatory roles in response to cold stress. In this study, we conducted
Quantitative Phosphoproteomics of the ATM and ATR dependent DNA damage response in Arabidopsis thaliana
Roitinger E, et al.
Molecular and Cellular Proteomics, 3(10), mcp-M114 (2015)
Jason A Wojcechowskyj et al.
PloS one, 6(9), e24918-e24918 (2011-09-29)
CXCL12 (SDF-1) is a chemokine that binds to and signals through the seven transmembrane receptor CXCR4. The CXCL12/CXCR4 signaling axis has been implicated in both cancer metastases and human immunodeficiency virus type 1 (HIV-1) infection and a more complete understanding
The SCX/IMAC enrichment approach for global phosphorylation analysis by mass spectrometry
Villen J and Gygi SP
Nature Protocols, 3(10), 1630-1630 (2008)

Articles

Protein modifications are crucial for disease study. Analysis methods are key.

Protein modifications are crucial for disease study. Analysis methods are key.

Protein modifications are crucial for disease study. Analysis methods are key.

Protein modifications are crucial for disease study. Analysis methods are key.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service