Skip to Content
Merck

E3528

EZview Red HIS-Select® HC Nickel Affinity Gel

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106500
NACRES:
NA.56
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


storage temp.

−20°C

Quality Segment

Application

Small-scale affinity capture of fusion recombinant proteins with His-tags / histidine-containing tags (His-tagged proteins).
When performing small scale affinity capture, such as immunoprecipitation, the affinity matrix is difficult to see in the microcentrifuge tubes. Accidental aspiration of the resin leads to quantitative variability in results. The EZview Red Affinity Gels greatly reduces the risk of pellet loss. EZview resins perform as well as conventional non-colored affinity gels, but allow the user to easily differentiate pellet from supernatant. This correlates to more accurate data because less protein is lost.

Features and Benefits

  • Increased visibility - Red color reduces risk of incidental aspiration
  • Improved recovery of target protein by reduced accidental loss
  • Higher reproducibility - More consistent yields

Physical form

1:1 (v/v) suspension in PBS containing 50% glycerol and MIT-HClis present at 30 PPM and 0.003% (w/v).

Legal Information

EZview is a trademark of Sigma-Aldrich Co. LLC
HIS-Select is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library