Skip to Content
Merck
All Photos(2)

Key Documents

I0383

Sigma-Aldrich

Isobutyryl coenzyme A lithium salt

≥85%

Synonym(s):

2-Methylpropanoyl-CoA, IB-CoA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H42N7O17P3S · xLi+
CAS Number:
Molecular Weight:
837.62 (free acid basis)
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Assay

≥85%

storage temp.

−20°C

SMILES string

[Li].CC(C)C(=O)SCCNC(=O)CCNC(=O)C(O)C(C)(C)COP(O)(=O)OP(O)(=O)OCC1OC(C(O)C1OP(O)(O)=O)n2cnc3c(N)ncnc23

InChI

1S/C25H42N7O17P3S.Li.H/c1-13(2)24(37)53-8-7-27-15(33)5-6-28-22(36)19(35)25(3,4)10-46-52(43,44)49-51(41,42)45-9-14-18(48-50(38,39)40)17(34)23(47-14)32-12-31-16-20(26)29-11-30-21(16)32;;/h11-14,17-19,23,34-35H,5-10H2,1-4H3,(H,27,33)(H,28,36)(H,41,42)(H,43,44)(H2,26,29,30)(H2,38,39,40);;

InChI key

XOJGPZXPXDNYHG-UHFFFAOYSA-N

Biochem/physiol Actions

Coenzyme A functions as an acyl group carrier, acetyl-CoA. Isobutyryl coenzyme A (IB-CoA), a short-chain branched acyl-CoA, provides the starter unit for biosynthesis of myxalamid B. Isobutyryl coenzyme A is used as a substrate to study the specificity and kinetics of isobutyryl-coenzyme A (CoA) mutase (EC 5.4.99.13).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helge B Bode et al.
Chembiochem : a European journal of chemical biology, 8(17), 2139-2144 (2007-10-24)
Myxalamids are potent inhibitors of the eukaryotic electron transport chain produced by different myxobacteria. Here, we describe the identification of the myxalamid biosynthesis gene cluster from Myxococcus xanthus. Additionally, new myxalamids (5-13) have been obtained by mutasynthesis from bkd mutants
Jinxia Lin et al.
Bioresource technology, 102(2), 1863-1868 (2010-10-29)
Supply of isobutanol to enhance the production of anti-tumor agent ansamitocin P-3 (AP-3) in medium containing agro-industrial residues was investigated with analysis of gene transcription, enzyme activity, and intermediate accumulation. Under the optimal addition of isobutanol, about 4-fold improvement of
A Ratnatilleke et al.
The Journal of biological chemistry, 274(44), 31679-31685 (1999-10-26)
Isobutyryl-CoA mutase (ICM) catalyzes the reversible, coenzyme B(12)-dependent rearrangement of isobutyryl-CoA to n-butyryl-CoA, which is similar to, but distinct from, that catalyzed by methylmalonyl-CoA mutase. ICM has been detected so far in a variety of aerobic and anaerobic bacteria, where
Mohini S Ghatge et al.
Applied and environmental microbiology, 75(11), 3469-3476 (2009-03-24)
Phoslactomycins (PLMs) and related leustroducsins (LSNs) have been isolated from a variety of bacteria based on antifungal, anticancer, and other biological assays. Streptomyces sp. strain HK 803 produces five PLM analogs (PLM A and PLMs C to F) in which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service