Skip to Content
Merck
All Photos(4)

Key Documents

A1625

Sigma-Aldrich

Acetoacetyl coenzyme A sodium salt hydrate

cofactor for acyl transfer

Synonym(s):

Acetoacetyl-CoANa3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H40N7O18P3S · xNa+ · yH2O
CAS Number:
Molecular Weight:
851.61 (anhydrous free acid basis)
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (Organic)

Assay

≥90%

form

powder

solubility

H2O: soluble~-50 g/L

storage temp.

−20°C

SMILES string

[Na+].[Na+].[Na+].CC(=O)CC(=O)SCCNC(=O)CCNC(=O)C(O)C(C)(C)COP([O-])(=O)OP(O)(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1OP([O-])([O-])=O)n2cnc3c(N)ncnc23

InChI

1S/C25H40N7O18P3S/c1-13(33)8-16(35)54-7-6-27-15(34)4-5-28-23(38)20(37)25(2,3)10-47-53(44,45)50-52(42,43)46-9-14-19(49-51(39,40)41)18(36)24(48-14)32-12-31-17-21(26)29-11-30-22(17)32/h11-12,14,18-20,24,36-37H,4-10H2,1-3H3,(H,27,34)(H,28,38)(H,42,43)(H,44,45)(H2,26,29,30)(H2,39,40,41)/t14-,18-,19-,20+,24-/m1/s1

InChI key

OJFDKHTZOUZBOS-CITAKDKDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Acetoacetyl coenzyme A (Acetoacetyl-CoA) may be used as a substrate for a number of enzymes including; acetoacetyl-CoA thiolase (EC 2.3.1) which produces acetyl-CoA in reverse condensation reaction and (3-Hydroxy-3-methylglutaryl coenzyme A (CoA) synthase (HMGCS)) which produces 3-hydroxy-3-methylglutaryl (HMG)-CoA in the mevalonate pathway leading to terpenoid biosynthesis. Acetoacetyl-CoA is also a precursor of poly-beta-hydroxybutyrate polymers in microorganisms.
Substrate for acetoacetyl-CoA thiolase EC 2.3.1.9; Role in the biosynthesis of nonactin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Werner J Kovacs et al.
Progress in lipid research, 41(5), 369-391 (2002-07-18)
Peroxisomes contain enzymes catalyzing a number of indispensable metabolic functions mainly related to lipid metabolism. The importance of peroxisomes in man is stressed by the existence of genetic disorders in which the biogenesis of the organelle is defective, leading to
James J Truglio et al.
The Journal of biological chemistry, 278(43), 42352-42360 (2003-08-12)
Bacterial enzymes of the menaquinone (Vitamin K2) pathway are potential drug targets because they lack human homologs. MenB, 1,4-dihydroxy-2-naphthoyl-CoA synthase, the fourth enzyme in the biosynthetic pathway leading from chorismate to menaquinone, catalyzes the conversion of O-succinylbenzoyl-CoA (OSB-CoA) to 1,4-dihydroxy-2-naphthoyl-CoA
Clementina Dellomonaco et al.
Nature, 476(7360), 355-359 (2011-08-13)
Advanced (long-chain) fuels and chemicals are generated from short-chain metabolic intermediates through pathways that require carbon-chain elongation. The condensation reactions mediating this carbon-carbon bond formation can be catalysed by enzymes from the thiolase superfamily, including β-ketoacyl-acyl-carrier protein (ACP) synthases, polyketide
Ethan I Lan et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(16), 6018-6023 (2012-04-05)
While conservation of ATP is often a desirable trait for microbial production of chemicals, we demonstrate that additional consumption of ATP may be beneficial to drive product formation in a nonnatural pathway. Although production of 1-butanol by the fermentative coenzyme
Rongcan Luo et al.
Signal transduction and targeted therapy, 6(1), 325-325 (2021-09-02)
Alzheimer's disease (AD) is characterized by progressive synaptic dysfunction, neuronal death, and brain atrophy, with amyloid-β (Aβ) plaque deposits and hyperphosphorylated tau neurofibrillary tangle accumulation in the brain tissue, which all lead to loss of cognitive function. Pathogenic mutations in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service