Skip to Content
Merck
All Photos(1)

Key Documents

H6132

Sigma-Aldrich

DL-3-Hydroxy-3-methylglutaryl coenzyme A sodium salt hydrate

≥90% (HPLC)

Synonym(s):

HMG-CoA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H42N7Na2O20P3S · xH2O
CAS Number:
Molecular Weight:
955.62 (anhydrous basis)
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Quality Level

Assay

≥90% (HPLC)

storage temp.

−20°C

SMILES string

O.[Na+].[Na+].CC(O)(CC(O)=O)CC(=O)SCCNC(=O)CCNC(=O)[C@@H](O)C(C)(C)COP(O)(=O)OP(O)(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1OP([O-])([O-])=O)n2cnc3c(N)ncnc23

InChI

1S/C27H44N7O20P3S.2Na.H2O/c1-26(2,21(40)24(41)30-5-4-15(35)29-6-7-58-17(38)9-27(3,42)8-16(36)37)11-51-57(48,49)54-56(46,47)50-10-14-20(53-55(43,44)45)19(39)25(52-14)34-13-33-18-22(28)31-12-32-23(18)34;;;/h12-14,19-21,25,39-40,42H,4-11H2,1-3H3,(H,29,35)(H,30,41)(H,36,37)(H,46,47)(H,48,49)(H2,28,31,32)(H2,43,44,45);;;1H2/q;2*+1;/p-2/t14-,19-,20-,21-,25-,27?;;;/m1.../s1

InChI key

QGWMCHPRNDWWMT-UAUHQYJPSA-L

General description

DL-3-Hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) is obtained from acetyl-CoA and acetoacetyl-CoA during condensation with the help of HMG-CoA synthase.

Application

DL-3-Hydroxy-3-methylglutaryl coenzyme A sodium salt hydrate has been used in drug treatment to study its effect on mTORC1 regulation. It has also been used to compare the acylomes of two reactive acyl-CoA species, like HMG-CoA and glutaryl-CoA.

Biochem/physiol Actions

DL-3-Hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) is a substrate used to study the specificity and kinetics of the enzyme 3-hydroxyl-3-methyl-glutaryl coenzyme A (HMG-CoA) reductase. HMG-CoA is the key intermediate in the biosynthsis of terpenes and ketone bodies. Its metabolism is the target of statin drugs used to control cholesterol levels.
DL-3-Hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) acts as a precursor of cholesterol synthesis. Inhibition of HMG-CoA can decrease cholesterol synthesis. This can be converted to β-hydroxybutyrate in the blood. HMG-CoA is a regulatory enzyme for cholesterol biosynthesis, it couples with LDL receptor to regulate cholesterol synthesis, once it is inhibited it would decrease cholesterol synthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alex Pitcher et al.
American heart journal, 169(5), 605-612 (2015-05-13)
A number of randomized trials are underway, which will address the effects of angiotensin receptor blockers (ARBs) on aortic root enlargement and a range of other end points in patients with Marfan syndrome. If individual participant data from these trials
Rajakrishnan Veluthakal et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 39(6), 2110-2120 (2016-11-02)
At least 300 prenylated proteins are identified in the human genome; the majority of which partake in a variety of cellular processes including growth, differentiation, cytoskeletal organization/dynamics and vesicle trafficking. Aberrant prenylation of proteins is implicated in human pathologies including
Distribution of Fatty Acids and Lipids During Pregnancy
Advances in Clinical Chemistry, 84, 209-239 (2018)
Yoshiko Mizuno et al.
Journal of atherosclerosis and thrombosis, 18(5), 351-358 (2011-03-24)
Atherosclerosis is a progressive disease causally associated with multiple cardiovascular risk factors, including dyslipidemia. Without effective intervention, atherosclerosis becomes evidenced clinically as coronary artery and cerebrovascular disease, both of which remain the leading causes of death worldwide. Multiple lines of
Psyllium Polysaccharide
Bioactive Polysaccharides, 395-443 (2018)

Articles

Cholesterol biosynthesis starts in the hepatic endoplasmic reticulum with acetyl-CoA, yielding 3-hydroxy-3-methylglutaryl-CoA via HMG-CoA synthase.

Cholesterol biosynthesis starts in the hepatic endoplasmic reticulum with acetyl-CoA, yielding 3-hydroxy-3-methylglutaryl-CoA via HMG-CoA synthase.

Cholesterol biosynthesis starts in the hepatic endoplasmic reticulum with acetyl-CoA, yielding 3-hydroxy-3-methylglutaryl-CoA via HMG-CoA synthase.

Cholesterol biosynthesis starts in the hepatic endoplasmic reticulum with acetyl-CoA, yielding 3-hydroxy-3-methylglutaryl-CoA via HMG-CoA synthase.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service