Skip to Content
Merck
All Photos(6)

Documents

HPA011311

Sigma-Aldrich

Anti-RPL10 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

RPL10 Antibody - Anti-RPL10 antibody produced in rabbit, Rpl10 Antibody, Anti-AC010422.7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, mouse, rat

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

KMSSCAGADRLQTGMRGAFGKPQGTVARVHIGQVIMSIRTKLQNKEHVIEALRRAKFKFPGRQKIHISKKWGFTKFNADEFEDMVAEKRLIPDGCGVKYIPSRGPLDKWW

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RPL10(6134)

General description

RPL10 (ribosomal protein L10) is a part of 60s ribosomal subunit, and is highly conserved in nature. It is localized to human chromosome Xq28. It is a ribosomal protein which belongs to L10e family. It is composed of 214 amino acids, and the core domain is made of amino acids 34-182. It has a two layered α-β structure, and its C-terminal resides in the outer surface of the ribosome. It is also a putative tumor suppressor gene.

Immunogen

QM protein homolog recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

RPL10 (ribosomal protein L10) is supposed to play an essential role in ribosomal assembly and function. It is incorporated in ribosome in the cytoplasm, and this incorporation is necessary for the initiation of translation. It interacts with nuclear-export adapter NMD3p, which acts as a nuclear export signal, and facilitates the transport of the ribosomal complex to the nucleus. Therefore, it plays a key role in the maturation of the ribosomal complex, and the synthesis of proteins. Studies show that mutations in this gene might be linked to premature ovarian failure. It acts as a transcription co-factor inhibiting the activity of AP (activating protein)-1 transcription factor. Studies show that down-regulation of RPL10 might be linked to the initial stages of prostate cancer. However, at later stages it is up-regulated and this might promote the development of an aggressive phenotype.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86616

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitsuhiro Nishimura et al.
Journal of molecular biology, 377(2), 421-430 (2008-02-09)
A phylogenetically conserved ribosomal protein L16p/L10e organizes the architecture of the aminoacyl tRNA binding site on the large ribosomal subunit. Eukaryotic L10 also exhibits a variety of cellular activities, and, in particular, human L10 is known as a putative tumor
Xiaohong Gong et al.
BMC medical genetics, 10, 7-7 (2009-01-27)
Autism spectrum disorders (ASD) are severe neurodevelopmental disorders with the male:female ratio of 4:1, implying the contribution of X chromosome genetic factors to the susceptibility of ASD. The ribosomal protein L10 (RPL10) gene, located on chromosome Xq28, codes for a
Ana Maria Massad-Costa et al.
Maturitas, 57(4), 399-404 (2007-06-15)
The aim of this study was to evaluate the presence of mutations in the coding region of the QM gene and fragile X in patients with premature ovarian failure and gonadal dysgenesis. After approval by the local Ethics Committee, blood
G Altinok et al.
Prostate cancer and prostatic diseases, 9(1), 77-82 (2005-12-07)
The QM protein is a transcription cofactor inhibiting the activity of AP-1 transcription factors and is also a ribosomal protein participating in protein synthesis. While protein synthesis is known to be increased in many cancers, inhibition of AP-1 activity presumably
Matthew L Kraushar et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(31), 10911-10926 (2015-08-08)
Neocortical development requires tightly controlled spatiotemporal gene expression. However, the mechanisms regulating ribosomal complexes and the timed specificity of neocortical mRNA translation are poorly understood. We show that active mRNA translation complexes (polysomes) contain ribosomal protein subsets that undergo dynamic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service