Skip to Content
Merck
All Photos(6)

Documents

HPA018126

Sigma-Aldrich

Anti-CAPRIN1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Caprin-1, Anti-Cell cycle associated protein 1, Anti-Cytoplasmic activation- and proliferation-associated protein 1, Anti-GPI-anchored membrane protein 1, Anti-GPI-anchored protein p137, Anti-GPI-p137, Anti-p137GPI

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000
western blot: 0.04-0.4 μg/mL

immunogen sequence

LWDLLEGKEKPVCGTTYKVLKEIVERVFQSNYFDSTHNHQNGLCEEEEAASAPAVEDQVPEAEPEPAEEYTEQSEVESTEYVNRQFMAETQFTSGEKEQVDEWTVETVEVVNSLQQQPQAASPSVPEPHSLTPVAQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CAPRIN1(4076)

General description

Cell cycle associated protein 1 (CAPRIN1) gene is mapped to human chromosome 11p13. The gene codes for highly conserved cytoplasmic phosphoprotein. CAPRIN1 belongs to the conserved caprin family. The protein is ubiquitously expressed and is characterized with two highly conserved regions, homologous region-1 and −2, arginine-glycine-glycine (RGG) motif and the RG enrichment region.

Immunogen

Caprin-1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-CAPRIN1 antibody produced in rabbit has been used in immunohistochemistry (IHC) and western blotting.

Biochem/physiol Actions

Cell cycle associated protein 1 (CAPRIN1) is associated with cell proliferation and is essential for normal advancement through the G1-S phase of the cell cycle. It is involved in the formation of stress granules via phosphorylation of eIF2α (a transcription initiation factor) and also regulates the translation of mRNAs of proteins related to synaptic plasticity in neurons. Studies have shown that Japanese encephalitis virus (JEV) causes a translational shutoff by preventing the formation of stress granules through interaction of JEV core protein with CAPRIN1. It has also been shown that it may be involved in the development of osteosarcoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72806

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yao Zhang et al.
American journal of cancer research, 8(10), 2116-2129 (2018-11-13)
MicroRNAs (miRNAs) have been reported to play an essential role in tumor development and progression. However, the function of miR-621 in hepatocellular carcinoma (HCC) remains largely unexplored. In this study, we found that miR-621 was downregulated in the HCC specimens
Role of xn-caprin1-j16c in carcinogenesis (Review)
Yang ZS, et al.
Oncology Letters, 18(12), 15-21 (2019)
Alice C Copsey et al.
The Biochemical journal, 474(18), 3109-3120 (2017-07-25)
DDX3X, a helicase, can interact directly with mRNA and translation initiation factors, regulating the selective translation of mRNAs that contain a structured 5' untranslated region. This activity modulates the expression of mRNAs controlling cell cycle progression and mRNAs regulating actin
Adam A Sabile et al.
Biochimica et biophysica acta, 1832(8), 1173-1182 (2013-03-27)
Osteosarcoma (OS) is the most common primary bone malignancy in children and adolescents. More than 30% of patients develop lung metastasis, which is the leading cause of mortality. Recently, the extracellular matrix protein Cyr61 has been recognized as a malignancy
Upregulation of caprin1 expression is associated with poor prognosis in hepatocellular carcinoma
Tan N, et al.
Pathology Research and Practice, 213(12), 1563-1567 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service