Skip to Content
Merck

Skip To

C7698

Cycloheximide

from microbial, ≥94% (TLC)

Synonym(s):

3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide, Actidione, Naramycin A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C15H23NO4
CAS Number:
Molecular Weight:
281.35
UNSPSC Code:
12352111
NACRES:
NA.77
PubChem Substance ID:
EC Number:
200-636-0
Beilstein/REAXYS Number:
88868
MDL number:
Assay:
≥94% (TLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

microbial

Quality Segment

assay

≥94% (TLC)

form

powder

color

white to off-white

solubility

ethanol: soluble, clear to hazy

antibiotic activity spectrum

fungi, yeast

mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

[H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC2CC(=O)NC(=O)C2

InChI

1S/C15H23NO4/c1-8-3-9(2)15(20)11(4-8)12(17)5-10-6-13(18)16-14(19)7-10/h8-12,17H,3-7H2,1-2H3,(H,16,18,19)/t8-,9-,11-,12+/m0/s1

InChI key

YPHMISFOHDHNIV-FSZOTQKASA-N

Gene Information

General description

Cycloheximide, also known as Actidione, is a glutarimide antibiotic commonly derived from the bacterium Streptomyces griseus. It acts as a potent inhibitor of protein biosynthesis in eukaryotic cells by disrupting the translocation step during translation, effectively blocking translational elongation. In microbiology, it plays a crucial role as a selection agent for resistant strains of yeast and fungi, proving invaluable in controlled experimental environments.

In cell biology and biochemical research, Cycloheximide showcases a dual nature concerning Apoptosis Induction, inducing or inhibiting apoptosis depending on the cell type. Its rapid and reversible effects make it an ideal choice for studying cellular processes and determining protein half-life. Cycloheximide also finds extensive applications in biomedical research, where it inhibits protein synthesis in eukaryotic cells studied in vitro (outside of organisms) and its effects are rapidly reversed by simply removing it. This makes Cycloheximide a go-to choice for exploring cell biology, biomedical and biochemical research, offering precise control and versatility in experiments.

Application

  • In yeast strains, cycloheximide has been used as a protein synthesis inhibitor in the cycloheximide chase experiment.
  • It has been used to inhibit translation in mammalian cells.
  • It has been used to suppress fungal growth.

Biochem/physiol Actions

Mode of Action: Translation inhibition in eukaryotes resulting in cell growth arrest and cell death. CHX is widely used for the selection of CHX-resistant strains of yeast and fungi, controlled inhibition of protein synthesis for detection of short-lived proteins and super-induction of protein expression, and apoptosis induction or facilitation of apoptosis induction by death receptors.

Activity Spectrum: Active against yeast and fungi like Candida, Aspergillus, Saccharomyces, Penicillium

Features and Benefits

  • High-quality antibiotic suitable for multiple research applications
  • Commonly used in Cell Biology and Biochemical applications

Preparation Note

Tightly closed. Dry. Keep in a well-ventilated place. Keep locked up or in an area accessible
only to qualified or authorized persons.

Other Notes

For additional information on our range of Biochemicals, please complete this form.
Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.


Still not finding the right product?

Explore all of our products under Cycloheximide


signalword

Danger

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Chronic 2 - Muta. 2 - Repr. 1B

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters