Skip to Content
Merck
All Photos(1)

Key Documents

H4415

Sigma-Aldrich

HC-030031

≥98% (HPLC), powder

Synonym(s):

1,2,3,6-Tetrahydro-1,3-dimethyl-N-[4-(1-methylethyl)phenyl]-2,6-dioxo-7H-purine-7-acetamide, 2-(1,3-Dimethyl-2,6-dioxo-1,2,3,6-tetrahydro-7H-purin-7-yl)-N-(4-isopropylphenyl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21N5O3
CAS Number:
Molecular Weight:
355.39
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

room temp

SMILES string

CC(C)c1ccc(NC(=O)Cn2cnc3N(C)C(=O)N(C)C(=O)c23)cc1

InChI

1S/C18H21N5O3/c1-11(2)12-5-7-13(8-6-12)20-14(24)9-23-10-19-16-15(23)17(25)22(4)18(26)21(16)3/h5-8,10-11H,9H2,1-4H3,(H,20,24)

InChI key

HEQDZPHDVAOBLN-UHFFFAOYSA-N

Biochem/physiol Actions

HC-030031 is a selective TRPA1 channel blocker, antagonizing formalin-evoked calcium influx and formalin-induced pain. IC50 is 5 μM. TRPA1 antagonism is a promising therapeutic target for chronic inflammatory or neurogenic pain.
HC-030031 is a selective TRPA1 channel blocker.
HC-030031 is a selective TRPA1 channel blocker. HC-030031 antagonizes formalin-evoked calcium influx. IC50 is 5 μM.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luana Fischer et al.
Inflammation research : official journal of the European Histamine Research Society ... [et al.], 66(4), 311-322 (2016-12-03)
Although TRPA1, SP, histamine and 5-hydroxytryptamine (5-HT) have recognized contribution to nociceptive mechanisms, little is known about how they interact with each other to mediate inflammatory pain in vivo. In this study we evaluated whether TRPA1, SP, histamine and 5-HT
Coline Legrand et al.
Scientific reports, 10(1), 11238-11238 (2020-07-10)
The transient receptor potential (TRP) channels family are cationic channels involved in various physiological processes as pain, inflammation, metabolism, swallowing function, gut motility, thermoregulation or adipogenesis. In the oral cavity, TRP channels are involved in chemesthesis, the sensory chemical transduction
Yan Xu et al.
The Turkish journal of gastroenterology : the official journal of Turkish Society of Gastroenterology, 29(6), 692-700 (2018-11-02)
Transient receptor potential ankyrin 1 (TRPA1) and substance P (SP), both expression in sensory neurons, have important roles in stress-induced duodenal lesions. The possible contribution of TRPA1 and SP to stress-induced duodenal lesions was explored by using the water immersion
Anthony Bosson et al.
Molecular neurodegeneration, 12(1), 53-53 (2017-07-08)
Excessive synaptic loss is thought to be one of the earliest events in Alzheimer's disease (AD). However, the key mechanisms that maintain plasticity of synapses during adulthood or initiate synapse dysfunction in AD remain unknown. Recent studies suggest that astrocytes
Florian Schillers et al.
The Journal of pharmacology and experimental therapeutics, 359(1), 18-25 (2016-07-28)
Propacetamol (PPCM) is a prodrug of paracetamol (PCM), which was generated to increase water solubility of PCM for intravenous delivery. PPCM is rapidly hydrolyzed by plasma esterases to PCM and diethylglycine and shares some structural and metabolic properties with lidocaine.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service