Skip to Content
Merck
All Photos(1)

Key Documents

SML0085

Sigma-Aldrich

A-967079

≥98% (HPLC)

Synonym(s):

(1E,3E)-1-(4-Fluorophenyl)-2-methyl-1-penten-3-one oxime

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14FNO
CAS Number:
Molecular Weight:
207.24
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥12 mg/mL

originator

Abbott

storage temp.

2-8°C

SMILES string

CCC(=N/O)\C(C)=C\c1ccc(F)cc1

InChI

1S/C12H14FNO/c1-3-12(14-15)9(2)8-10-4-6-11(13)7-5-10/h4-8,15H,3H2,1-2H3/b9-8+,14-12+

InChI key

HKROEBDHHKMNBZ-CHBKHGQFSA-N

General description

A-967079 prevents neuropathic and inflammatory pain. It reduces cold allodynia, which is produced by nerve injury.

Application

A-967079 has been used in blocking miR-711-induced TRPA1 (transient receptor potential cation channel subfamily A member 1) channel.

Biochem/physiol Actions

A-967079 is a potent and selective antagonist of Transient Receptor Potential Anykrin 1 (TRPA1) with IC50′s of 67 nM and 289 nM at human and rat TRPA1 receptors, respectively, and minimal or no activity at other TRP channels or G-protein-coupled receptors, enzymes, transporters, and ion channels out of 89 tested. A-967079 blocks TRPA1 activation in human and rat cell lines and has been shown to reduce the responses of wide dynamic range (WDR) and nociceptive specific (NS) neurons to high-intensity mechanical stimulation.
TRPA1 receptor antagonist

Features and Benefits

This compound was developed by Abbott. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

miRNA-711 binds and activates TRPA1 extracellularly to evoke acute and chronic pruritus
Han Q, et al.
Neuron, 99(3), 449-463 (2018)
Molecular basis determining inhibition/activation of nociceptive receptor TRPA1: a single amino acid dictates species-specific actions of the most potent mammalian trpa1 antagonists
Banzawa N, et al.
Test, jbc-M114 (2014)
Molecular basis determining inhibition/activation of nociceptive receptor TRPA1: a single amino acid dictates species-specific actions of the most potent mammalian trpa1 antagonists
Banzawa N, et al.
The Journal of biological chemistry, jbc-M114 (2014)
Molecular basis determining inhibition/activation of nociceptive receptor TRPA1: a single amino acid dictates species-specific actions of the most potent mammalian trpa1 antagonists
Banzawa N, et al.
The Journal of Biological Chemistry, jbc-M114 (2014)
Steve McGaraughty et al.
Molecular pain, 6, 14-14 (2010-03-09)
There is growing evidence supporting a role for TRPA1 receptors in the neurotransmission of peripheral mechanical stimulation. In order to enhance understanding of TRPA1 contributions to mechanotransmission, we examined the effects a selective TRPA1 receptor antagonist, A-967079, on spinal neuronal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service