Skip to Content
Merck

SML1467

ASP7663

≥98% (HPLC)

Synonym(s):

(2E)-2-[7-Fluoro-1,2-dihydro-1-(2-methylpropyl)-2-oxo-3H-indol-3-ylidene]acetic acid, (E)-[7-Fluoro-1-(2-methylpropyl)-2-oxo-1,2-dihydro-3H-indol-3-ylidene]acetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H14FNO3
CAS Number:
Molecular Weight:
263.26
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

light orange to dark orange

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC(C)CN(C1=C(F)C=CC=C1/C2=C\C(O)=O)C2=O

InChI

1S/C14H14FNO3/c1-8(2)7-16-13-9(4-3-5-11(13)15)10(14(16)19)6-12(17)18/h3-6,8H,7H2,1-2H3,(H,17,18)/b10-6+

InChI key

RCVZUIGCNAAMIC-UXBLZVDNSA-N

Biochem/physiol Actions

ASP7663 is a selective activator of transient receptor potential ankyrin 1 (TRPA1), known to play a role for chemical sensing in the pain pathway. ASP7663 activated human, rat, and mouse TRPA1 receptors with EC50 values of 510, 540, and 500 nM, respectively, and little or no affinity at more than 60 other related receptors, channels and enzymes. ASP7663 enhanced colonic motility and exerted analgesic effects against abdominal pain in mouse models.
ASP7663 is a selective activator of transient receptor potential ankyrin 1 (TRPA1).


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library