Skip to Content
Merck
All Photos(1)

Key Documents

GE17-0756-01

Glutathione Sepharose 4B

Cytiva 17-0756-01, pack of 10 mL

Synonym(s):

affinity chromatography resin, GST-tagged proteins purification,

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

ligand

glutathione

packaging

pack of 10 mL

manufacturer/tradename

Cytiva 17-0756-01

matrix

4% agarose

particle size

45-165 μm

average diameter

90 μm

cleaning in place

4-13

working range

4-13

capacity

>25 mg binding capacity (horse liver GST/ml medium)

Related Categories

General description

Binding of GST to glutathione is flow-rate dependent and lower flow rates often increase the binding capacity. This is important to consider during sample loading and elution.
Glutathione Sepharose 4B is designed for high capacity single-step purification of Glutathione S-Transferase (GST) tagged fusion proteins.

GST-tagged proteins and other glutathione S-transferases or glutathone-dependent proteins can be purified directly from bacterial lysates using Glutathione Sepharose 4B. Proteins are eluted under mild, non-denaturing conditions that preserve protein antigenicity.

Features and Benefits

  • High capacity single step purification of recombinant glutathione S-transferase (GST) tagged proteins
  • Proteins are eluted under mild, non-denaturing conditions that preserve protein antigenicity and functionality.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin Zhou et al.
Nature communications, 11(1), 2674-2674 (2020-05-31)
Increasing evidence indicates that guidance molecules used during development for cellular and axonal navigation also play roles in synapse maturation and homeostasis. In C. elegans the netrin receptor UNC-40/DCC controls the growth of dendritic-like muscle cell extensions towards motoneurons and
Gerard H Pieper et al.
Developmental cell, 53(1), 27-41 (2020-02-29)
Eukaryotic genomes are organized within the nucleus through interactions with inner nuclear membrane (INM) proteins. How chromatin tethering to the INM is controlled in interphase and how this process contributes to subsequent mitotic nuclear envelope (NE) remodeling remains unclear. We have
Amandine Cartier-Michaud et al.
PLoS genetics, 13(6), e1006803-e1006803 (2017-06-16)
Spermatogenesis is a dynamic process that is regulated by adhesive interactions between germ and Sertoli cells. Germ cells express the Junctional Adhesion Molecule-C (JAM-C, encoded by Jam3), which localizes to germ/Sertoli cell contacts. JAM-C is involved in germ cell polarity
Ye Guan et al.
eLife, 7 (2018-04-06)
Single-cell analysis is pivotal to deciphering complex phenomena like heterogeneity, bistability, and asynchronous oscillations, where a population ensemble cannot represent individual behaviors. Bulk cell-free systems, despite having unique advantages of manipulation and characterization of biochemical networks, lack the essential single-cell
Amandine Draime et al.
Biochimica et biophysica acta, 1861(5), 534-542 (2018-02-23)
Understanding how the activity of transcription factors like HOX proteins is regulated remains a widely open question. In a recent screen for proteins interacting with HOXA1, we identified a PRDM protein family member, PRDM14, which is known to be transiently

Articles

This page addresses common problems associated with the expression and growth of GST-tagged proteins using Cytiva products.

This page shows how to purify GST-tagged proteins using GST SpinTrap™ from Cytiva.

This page describes general considerations for purification of GST-tagged proteins using Cytiva products.

GST-tagged protein yield varies based on protein, host cell, and culture conditions, influencing production.

See All

Protocols

This page describes enzymatic detection of GST-tagged proteins with the GST Detection Module from Cytiva.

This page describes enzymatic detection of GST-tagged proteins with the GST Detection Module from Cytiva.

This page describes enzymatic detection of GST-tagged proteins with the GST Detection Module from Cytiva.

This page describes enzymatic detection of GST-tagged proteins with the GST Detection Module from Cytiva.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service