Skip to Content
Merck
All Photos(1)

Key Documents

G9652

Sigma-Aldrich

Monosialoganglioside GM1 from bovine brain

lyophilized powder, BioXtra, γ-irradiated, ≥95% (TLC)

Synonym(s):

Ganglioside GM1, monosialo

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.75

Quality Level

sterility

γ-irradiated

product line

BioXtra

Assay

≥95% (TLC)

form

lyophilized powder

mol wt

~1540

technique(s)

blocking: suitable

impurities

endotoxin, tested

solubility

chloroform/methanol: 9.80-10.20 mg/mL, clear to slightly hazy, colorless to light yellow

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCCCC(=O)N[C@H](CO[C@@H]1O[C@H](CO)[C@@H](O[C@@H]2O[C@H](CO)[C@H](O[C@@H]3O[C@H](CO)[C@H](O)[C@H](O[C@@H]4O[C@H](CO)[C@H](O)[C@H](O)[C@H]4O)[C@H]3NC(C)=O)[C@H](O[C@@]5(C[C@H](O)[C@@H](NC(C)=O)[C@@H](O5)[C@H](O)[C@H](O)CO)C(O)=O)[C@H]2O)[C@H](O)[C@H]1O)[C@@H](O)\C=C\CCCCCCCCCCCCCCC

InChI

1S/C73H131N3O31/c1-5-7-9-11-13-15-17-19-20-22-24-26-28-30-32-34-52(86)76-44(45(83)33-31-29-27-25-23-21-18-16-14-12-10-8-6-2)40-99-69-61(93)59(91)63(50(38-79)102-69)104-71-62(94)67(98-41-47(85)55(87)66-53(74-42(3)81)46(84)35-73(97,107-66)72(95)96)64(51(39-80)103-71)105-68-54(75-43(4)82)65(57(89)49(37-78)100-68)106-70-60(92)58(90)56(88)48(36-77)101-70/h31,33,44-51,53-71,77-80,83-85,87-94,97H,5-30,32,34-41H2,1-4H3,(H,74,81)(H,75,82)(H,76,86)(H,95,96)/b33-31+/t44?,45?,46-,47?,48+,49+,50+,51+,53+,54+,55?,56-,57-,58-,59+,60+,61+,62+,63+,64-,65+,66+,67+,68-,69+,70-,71-,73-/m0/s1

InChI key

CNLVNZJJSHZYAS-ALSOZVJRSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Cer-Glc-Gal(NeuAc)-GalNAc-Gal

General description

Gangliosides are major constituents of neuronal cell membranes and endoplasmic reticulum. They contain a sialated polysaccharide chain linked to ceramide through a β-glycosidic linkage.
Gangliosides are major constituents of neuronal cell membranes and endoplasmic reticulum. They contain a sialated polysaccharide chain linked to ceramide through a β-glycosidic linkage. For classification of gangliosides see Svennerholm, L., et al. (eds.), Structure and Function of Gangliosides, New York, Plenum, 1980.

Application

Specific receptors for cholera toxin that accumulate in the brain in late infantile lipidosis.

Biochem/physiol Actions

Monosialoganglioside GM1 is a major sialoglycolipid of neuronal membranes that modulates calcium homeostasis. It binds to cholera toxin B subunit, resulting in stimulation of adenylate cyclase in a wide variety of cell types. After cholera toxin binds to membrane associated monosialoganglioside GM1, the A subunit of cholera toxin is translocated to the cell interior, where it catalyzes the ADP ribosylation of the membrane associated Gs subunit of adenylate cyclase. In addition, binding of cholera toxin to monosialoganglioside GM1 causes translocation of NF-κB and activation of dendritic cells.

Monosialoganglioside GM1 was one of many mono- and oligosaccharide ligands studied for their affinity for NKR-P1, a membrane protein on natural killer cells, which contains an extracellular Ca2+-dependent lectin domain. Monosialoganglioside GM1 is effective in partially correcting the consequences of neuroinjury in a number of in vivo and in vitro model systems.

Preparation Note

Monosialoganglioside GM1 is prepared by a modification of a published procedure. It is isolated from bovine brain by extraction and solvent fractionation methods. The final purification is done by HPLC on silica gel allowing no buffer salts to be used for the purification.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tobias Gustafsson et al.
European journal of immunology, 43(7), 1779-1788 (2013-05-08)
Cholera toxin (CT) binds to GM1-ganglioside receptors present on all nucleated cells. Despite this, it is a very potent mucosal adjuvant that has a dramatic impact on immune cells, as well as nerve and epithelial cells, causing diarrhea. This fact
Motonari Nomura et al.
Cancer science, 104(2), 238-244 (2012-11-09)
Hemagglutinating virus of Japan-envelope (HVJ-E) is a drug delivery vector based on inactivated Sendai virus. Recently, antitumor activities were found for HVJ-E itself and clinical trials of HVJ-E for some malignant tumors are now ongoing. We investigated the in vitro
Michael G Jobling et al.
mBio, 3(6), doi:10-doi:10 (2012-11-01)
Cholera toxin (CT) from Vibrio cholerae is responsible for the majority of the symptoms of the diarrheal disease cholera. CT is a heterohexameric protein complex with a 240-residue A subunit and a pentameric B subunit of identical 103-residue B polypeptides.
Eduardo Nobile-Orazio et al.
Journal of neurology, neurosurgery, and psychiatry, 85(7), 754-758 (2013-08-03)
Increased titres of serum IgM antibodies to GM1 ganglioside are often associated with multifocal motor neuropathy (MMN). Testing for IgM antibodies to other antigens including GM2, the mixture of GM1 and galactocerebroside (GM1/GalC) and the disulfated heparin disaccharide NS6S were
Djurre H de Jong et al.
Faraday discussions, 161, 347-363 (2013-06-29)
We present results from coarse grain molecular dynamics simulations of mixed model membranes consisting of saturated and unsaturated lipids together with cholesterol, in which lipid-anchored membrane proteins are embedded. The membrane proteins studied are the peripherally bound H-Ras, N-Ras, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service