Skip to Content
Merck
All Photos(1)

Key Documents

G8146

Sigma-Aldrich

Disialoganglioside GD1b from bovine brain

~95%, lyophilized powder

Synonym(s):

Ganglioside GD1b, disialo

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

~95%

Quality Level

form

lyophilized powder

storage temp.

−20°C

InChI

1S/C80H144N4O37/c1-4-6-8-10-12-14-16-18-19-21-23-25-27-29-31-33-54(96)83-36-49(44(91)32-30-28-26-24-22-20-17-15-13-11-9-7-5-2)111-74-65(104)63(102)67(52(40-88)114-74)116-76-66(105)72(68(53(41-89)115-76)117-73-57(84-43(3)90)71(61(100)51(39-87)112-73)118-75-64(103)62(101)60(99)50(38-86)113-75)121-80(78(108)109)35-46(93)56(82)70(120-80)59(98)48(95)42-110-79(77(106)107)34-45(92)55(81)69(119-79)58(97)47(94)37-85/h30,32,44-53,55-76,85-89,91-95,97-105H,4-29,31,33-42,81-82H2,1-3H3,(H,83,96)(H,84,90)(H,106,107)(H,108,109)/b32-30+/t44?,45-,46+,47?,48?,49?,50+,51+,52-,53+,55+,56-,57+,58?,59?,60-,61-,62-,63-,64+,65-,66+,67-,68-,69+,70-,71+,72+,73-,74-,75-,76-,79-,80-/m1/s1

InChI key

AIUBMTQAHKTQMI-MPGPKOMUSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Cer-Glc-Gal(NeuAc-NeuAc)-GalNAc-Gal

General description

Gangliosides are major constituents of neuronal cell membranes and endoplasmic reticulum; contain a sialated polysaccharide chain linked to ceramide through a β-glycosidic linkage; for classification of gangliosides see Svennerholm, L., et al. (eds.), Structure and Function of Gangliosides, New York, Plenum, 1980.

Biochem/physiol Actions

May act as a bacterial toxin receptor (tetanus, botulinus); high levels found in gliomas and astrocytomas.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beatriz Valle-Argos et al.
Neuro-oncology, 12(11), 1135-1146 (2010-07-10)
In spite of their low incidence, central nervous system tumors have elevated morbidity and mortality, being responsible for 2.3% of total cancer deaths. The ganglioside O-acetylated GD1b (O-Ac GD1b; neurostatin), present in the mammalian brain, and the semi-synthetic O-butyrylated GD1b
Seong-Hae Jeong et al.
Journal of neuro-ophthalmology : the official journal of the North American Neuro-Ophthalmology Society, 31(4), 326-330 (2011-09-16)
Antiganglioside antibodies are found in various neurological disorders that constitute a continuum from peripheral neuropathy to encephalitis. However, nystagmus has rarely been described in patients with ataxia associated with antiganglioside antibodies. From January 2008 to July 2009, we identified 3
Alina Serb et al.
European journal of mass spectrometry (Chichester, England), 15(4), 541-553 (2009-08-08)
Gangliosides (GGs), a large group of sialylated glycosphingolipids, are considered biomarkers of human brain development, aging and certain diseases. Determination of individual GG components in complex mixtures extracted from a human brain represents a fundamental prerequisite for correlating their specificity
Ana L Moyano et al.
Glycobiology, 22(12), 1768-1774 (2012-07-31)
Immunization of rabbits with bovine brain gangliosides induced an experimental neuropathy, with clinical signs resembling Guillain-Barré syndrome. All the immunized animals developed immunoglobulin G immunoreactivity to GM1 ganglioside. In a few (4 of 27) animals, an additional anti-ganglioside antibody population
Keiichiro Susuki et al.
Experimental neurology, 233(1), 534-542 (2011-12-20)
Autoantibodies against gangliosides GM1 or GD1a are associated with acute motor axonal neuropathy (AMAN) and acute motor-sensory axonal neuropathy (AMSAN), whereas antibodies to GD1b ganglioside are detected in acute sensory ataxic neuropathy (ASAN). These neuropathies have been proposed to be

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service