Skip to Content
Merck
All Photos(5)

Key Documents

G6654

Sigma-Aldrich

L-Glutathione oxidized

≥98%, BioXtra

Synonym(s):

GSSG, Glutathiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32N6O12S2
CAS Number:
Molecular Weight:
612.63
Beilstein:
1718700
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

L-Glutathione oxidized, BioXtra, ≥98%

product line

BioXtra

Quality Level

Assay

≥98%

form

powder

impurities

≤0.005% Phosphorus (P)
≤0.1% Insoluble matter
≤6% ethanol

ign. residue

≤0.5%

color

white

solubility

H2O: 0.1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.001%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

storage temp.

2-8°C

SMILES string

N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O

InChI

1S/C20H32N6O12S2/c21-9(19(35)36)1-3-13(27)25-11(17(33)23-5-15(29)30)7-39-40-8-12(18(34)24-6-16(31)32)26-14(28)4-2-10(22)20(37)38/h9-12H,1-8,21-22H2,(H,23,33)(H,24,34)(H,25,27)(H,26,28)(H,29,30)(H,31,32)(H,35,36)(H,37,38)/t9-,10-,11-,12-/m0/s1

InChI key

YPZRWBKMTBYPTK-BJDJZHNGSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Glu-Cys-Gly, Glu-Cys-Gly [Disulfide bridge: 2a-2b]

Biochem/physiol Actions

L-Glutathione, oxidized is frequently measure in vivo as an indicator of oxidative stress. Oxidized L-Glutathione may be converted to L-glutathione by various reducing systems.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mary L Bath
PloS one, 5(2), e9387-e9387 (2010-03-03)
In vitro fertilization (IVF) of eggs by frozen and thawed C57BL/6J mouse sperm is inhibited by dead sperm and enhanced by preincubation of the sperm in calcium-free medium. In other species, the presence of sperm killed by freezing and thawing
N O Kaakoush et al.
Antimicrobial agents and chemotherapy, 53(5), 1884-1891 (2009-02-19)
Metronidazole resistance in Helicobacter pylori has been attributed to mutations in rdxA or frxA. Insufficient data correlating RdxA and/or FrxA with the resistant phenotype, and the emergence of resistant strains with no mutations in either rdxA or frxA, indicated that
S K Tandon et al.
Toxicology letters, 145(3), 211-217 (2003-10-29)
The influence of an antioxidant agent such as N-acetyl cysteine (NAC) or mannitol on the cadmium chelating ability of monoisoamyl 2,3-dimercaptosuccinate (MiADMS) was investigated in cadmium pre-exposed rats. This ester of 2,3-dimercaptosuccinic acid (DMSA), an accepted drug for lead poisoning
P Aukrust et al.
Blood, 86(1), 258-267 (1995-07-01)
We investigated the intracellular glutathione redox status in isolated lymphocyte subpopulations and monocytes in patients with human immunodeficiency virus type 1 (HIV-1) infection and in healthy controls. CD4+ lymphocytes from HIV-1-infected patients were primarily characterized by a substantial increase in
T Yoshida
Journal of chromatography. B, Biomedical applications, 678(2), 157-164 (1996-04-12)
A method is described for simultaneous quantitation of reduced (GSH) and oxidized (GSSG) glutathione in erythrocytes by HPLC. They were determined by standard addition method. Blood samples were collected in tubes containing 1,10-phenanthroline. The separated erythrocytes were hemolyzed with water

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service