Skip to Content
Merck

3541

Glutathione, Reduced, Free Acid

>98% (titration), solid, endogenous antioxidant, Calbiochem®

Synonym(s):

Glutathione, Reduced, Free Acid, γ-Glu-Cys-Gly, GSH

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H17N3O6S
CAS Number:
Molecular Weight:
307.32
UNSPSC Code:
12352209
NACRES:
NA.25
MDL number:
Assay:
>98% (titration)
Form:
solid
Quality level:
Storage condition:
OK to freeze, desiccated (hygroscopic)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Glutathione, Reduced, Free Acid, Glutathione, Reduced, Free Acid, CAS 70-18-8, is a tripeptide that serves as an endogenous antioxidant and provides protection against auto-oxidation.

Quality Level

description

RTECS - MC0556000
Merck USA index - 14, 4475

assay

>98% (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic)

color

white

solubility

water: 1 mg/mL, DMF: 10 mg/mL, ethanol: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

SCC(NC(=O)CCC([N+H3])C(=O)[O-])C(=O)NCC(=O)O

InChI

1S/C10H17N3O6S/c11-5(10(18)19)1-2-7(14)13-6(4-20)9(17)12-3-8(15)16/h5-6,20H,1-4,11H2,(H,12,17)(H,13,14)(H,15,16)(H,18,19)

InChI key

RWSXRVCMGQZWBV-UHFFFAOYSA-N

General description

Glutathione is a tripeptide composed of cysteine, glycine, and glutamic acid. In cells glutathione either exists as reduced (GSH) and oxidized (GSSG). The ratio of glutathione reduced (GSH) to glutathione oxidized (GSSG) determines the cell’s redox status. Healthy resting cells have a GSH/GSSG ratio >100. In cells under oxidative stress, the GSH/GSSG ratio drops to between 1 and 10. Glutathione has also been identified as the thiol buffer that keeps sulfohydryl groups in the reduced form of many proteins. It is produced solely in the cytochrome P450 system and actively pumped to mitochondria. GSH depletion can lead to Neurodegenerative disorders, Pulmonary disease, Immune diseases, Cardiovascular diseases, Chronic age-related diseases, Liver disease, Cystic fibrosis and Aging process itself.

Application

Glutathione, Reduced, Free Acid has been used to elute the GST-OTUD6A proteins after purification using Glutathione Sepharose.

Biochem/physiol Actions

Cell permeable: no
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

"Aging: getting older, exhibiting the signs of age, the decline in the physical (and mental) well-being over time, leading to death. Since the beginning of time, man has been obsessed with trying to slow down, stop, or even reverse the signs of aging. Many have gone as far as experimenting with nutritional regimens, eccentric exercises, fantastic rituals, and naturally occurring or synthetic wonder-elements to evade the signs of normal aging. Biologically speaking, what is aging? And what does the latest research tell us about the possibility of discovering the elusive “fountain of youth”? Many advances in our understanding of aging have come from systematic scientific research, and perhaps it holds the key to immortality. Scientifically, aging can be defined as a systems-wide decline in organismal function that occurs over time. This decline occurs as a result of numerous events in the organism, and these events can be classified into nine “hallmarks” of aging, as proposed by López-Otin et al. (2013). Several of the pathologies associated with aging are a direct result of these events going to extremes and may also involve aberrant activation of proliferation signals or hyperactivity. The hallmarks of aging have been defined based on their fulfillment of specific aging related criteria, such as manifestation during normal aging, acceleration of aging if experimentally induced or aggravated, and retardation of aging if prevented or blocked, resulting in increased lifespan. The nine hallmarks of aging are genomic instability, telomere attrition, epigenetic alterations, loss of proteostasis, deregulated nutrient sensing, mitochondrial dysfunction, cellular senescence, stem cell exhaustion, and altered intercellular communication. The biological processes underlying aging are complex. By understanding the hallmarks in greater detail, we can get closer to developing intervention strategies that can make the aging process less of a decline, and more of a recline."

Global Trade Item Number

SKUGTIN
PHR2586-1G04061842563999
RDD017-1KG04061836828547
4005-500GM04055977241945
03620-250G04061838619020
ED-100G04061835544721
ED-5KG04061833620076
EDS-100G04061835544851
E6758-100G04061835544592
E8008-100ML04061838259424
E9884-1KG04061835544707
03620-1KG04061838619013
798681-500G04061832973852
798681-1KG04061832973845
1233508-200MG04061838722256
E9884-100G04061835544691
E9884-500G04061835544714
EDS-1KG04061835544868

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service