Skip to Content
Merck
All Photos(1)

Documents

G5018

Sigma-Aldrich

Anti-Goat Serum antibody produced in rabbit

whole antiserum

Synonym(s):

Anti-Serum, goat

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

species reactivity

goat

technique(s)

immunoelectrophoresis: suitable

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Specificity

Strong reactivity with normal goat serum has been determined by immunoelectrophoresis (IEP). This antiserum has not been assayed for interspecies crossreactivity.

Application

Anti-Goat Serum antibody produced in rabbit has been used in immunochemistry.
Anti-Goat Serum antibody produced in rabbit was used as control in protein characterization of serum albumin using Ouchterlony gel diffusion method.

Biochem/physiol Actions

Serum antibodies have their significance in the innate and adaptive immune responses. They are closely associated with immune system related pathogenesis.

Physical form

Rabbit Anti-Goat Serum is provided as a liquid containing 0.1% sodium azide as a preservative.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J E Bassett et al.
Experimental physiology, 77(6), 839-848 (1992-11-01)
Intra-articular injection of radiolabelled, commercial goat serum albumin (GSA) produces acute arthritis in caprine joints. This inflammation distorts clearance values and vitiates studies of normal lymphatic function. Endotoxin, routinely found in commercial albumin preparations, appears to cause this local reaction.
Rudolf A Manz et al.
Annual review of immunology, 23, 367-386 (2005-03-18)
In vertebrates, serum antibodies are an essential component of innate and adaptive immunity and immunological memory. They also can contribute significantly to immunopathology. Their composition is the result of tightly regulated differentiation of B lymphocytes into antibody-secreting plasma blasts and
Host influence on the nutritional and reproductive status of Triatoma infestans (Klug)(Hemiptera: Reduviidae) peridomiciliary populations
Soria C, et al.
Revista de la Sociedad Entomologica Argentina, 78(2) (2019)
Lihui Wang et al.
STAR protocols, 2(3), 100620-100620 (2021-07-02)
Telomere dysfunction-induced focus (TIF) assay allows efficient profiling of telomere dysfunctions in cells and tissues. Here, we describe the use of the TIF assay to screen synthetic peptides from E3 ubiquitin ligase FBW7, a tumor suppressor gene product, to prevent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service