Skip to Content

B9398

Anti-Bovine Serum antibody produced in rabbit

whole antiserum

Synonym(s):

Anti-Serum, bovine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
MDL number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IEP, IF (d), IHC (p)
Citations:
3
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

species reactivity

bovine

technique(s)

direct immunofluorescence: 1:160 using human peripheral blood lymphocytes, immunoelectrophoresis: suitable, immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:160

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

bovine ... Alb(280717)

General description

Bovine serum consists of fats, plasma proteins, growth factors, peptides, carbohydrates, hormones and inorganic substances.

Application

Anti-Bovine Serum antibody produced in rabbit may be used to prepare anti-BSA-BSA complexes that enhance the relative procoagulant activity of leukocytes. It has been used in
  • immunoelectrophoresis
  • immunodiffusion
  • enzyme linked immunosorbent assay (ELISA)

Biochem/physiol Actions

Bovine serum is required for the in vitro growth of cells. Bovine serum albumin (BSA) is one of the important constituents of bovine serum.
Rabbit Anti-Bovine Whole Serum is determined to have strong reactivity with bovine serum proteins. No reactivity with horse or pig serum proteins is observed.

Physical form

Supplied as a liquid containing 0.1% sodium azide (see MSDS) as a preservative.

Preparation Note

For continuous use, store at 2-8 °C for up to one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Other Notes

Antiserum adsorbed with equine and porcine serum proteins

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library