Skip to Content
Merck

H3383

Anti-Human Serum antibody produced in rabbit

fractionated antiserum, lyophilized powder

Synonym(s):

Anti Human Serum, Anti Human Serum - Anti-Human Serum antibody produced in rabbit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
MDL number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IEP
Citations:
4
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

fractionated antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

packaging

vial of 2 mL lyophilized antiserum

technique(s)

immunoelectrophoresis: 1:10 using human serum

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Serum is the component of blood plasma without cells and clotting factors. It is used in various assays, research and also as a supplement in cell culture. Anti-Human Serum antibody can be used to study the response of antibody against intratracheally administered lipopolysaccharide . Rabbit anti-serum antibody reacts specifically with human serum.

Immunogen

Normal human serum

Application

Anti-Human Serum antibody produced in rabbit has been used for inner cell mass cluster (ICM) isolation in embryonic stem cells.
Anti-Human Serum antibody can be used in immunoelectrophoresis.

Physical form

Lyophilized from 0.01 M phosphate buffered saline, pH 7.2

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library